Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122801412-122801685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC62
Entrez Gene ID
84660
Gene Name
coiled-coil domain containing 62
Gene Aliases
CT109, ERAP75, TSP-NY
UniGene
Hs.592008
Gene Chromosome Location
Chr.12: 122773621-122827385 on Build GRCh38
Transcripts
NM_201435.4 NR_027918.2 XM_006719643.1 XM_006719644.1 XM_011538856.1 XM_011538857.1 XM_011538859.1 XM_017020033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3457400 c.1367C>T p.S456F Chr.12: 122801513-122801513 on Build GRCh38
3457398 c.1337G>A p.G446E Chr.12: 122801483-122801483 on Build GRCh38
7180438 c.1480G>A p.E494K Chr.12: 122801626-122801626 on Build GRCh38
6036504 c.1412C>G p.T471S Chr.12: 122801558-122801558 on Build GRCh38
6885464 c.1325T>C p.V442A Chr.12: 122801471-122801471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777184165 Chr.12: 122801562-122801562 on Build GRCh38
rs776798019 Chr.12: 122801648-122801648 on Build GRCh38
rs761358663 Chr.12: 122801586-122801586 on Build GRCh38
rs923962912 Chr.12: 122801654-122801654 on Build GRCh38
rs540867038 Chr.12: 122801459-122801459 on Build GRCh38
rs778790320 Chr.12: 122801505-122801505 on Build GRCh38
rs751097754 Chr.12: 122801456-122801456 on Build GRCh38
rs965023758 Chr.12: 122801548-122801548 on Build GRCh38
rs773618660 Chr.12: 122801534-122801534 on Build GRCh38
rs138955637 Chr.12: 122801564-122801564 on Build GRCh38
rs767122360 Chr.12: 122801597-122801597 on Build GRCh38
rs549775391 Chr.12: 122801596-122801596 on Build GRCh38
rs778931723 Chr.12: 122801621-122801621 on Build GRCh38
rs749239981 Chr.12: 122801492-122801492 on Build GRCh38
rs375118548 Chr.12: 122801664-122801664 on Build GRCh38
rs995721836 Chr.12: 122801466-122801466 on Build GRCh38
rs750997907 Chr.12: 122801579-122801579 on Build GRCh38
rs768650790 Chr.12: 122801504-122801504 on Build GRCh38
rs761090770 Chr.12: 122801553-122801553 on Build GRCh38
rs149451449 Chr.12: 122801568-122801568 on Build GRCh38
rs780033896 Chr.12: 122801479-122801479 on Build GRCh38
rs145950256 Chr.12: 122801622-122801622 on Build GRCh38
rs990690474 Chr.12: 122801656-122801656 on Build GRCh38
rs748216674 Chr.12: 122801625-122801625 on Build GRCh38
rs753671216 Chr.12: 122801611-122801611 on Build GRCh38
rs772319364 Chr.12: 122801626-122801626 on Build GRCh38
rs772372235 Chr.12: 122801533-122801533 on Build GRCh38
rs200798716 Chr.12: 122801601-122801601 on Build GRCh38
rs1019412609 Chr.12: 122801547-122801547 on Build GRCh38
rs775349370 Chr.12: 122801511-122801511 on Build GRCh38
rs755888041 Chr.12: 122801602-122801602 on Build GRCh38
rs763572618 Chr.12: 122801452-122801452 on Build GRCh38
rs569908909 Chr.12: 122801665-122801665 on Build GRCh38
rs771200702 Chr.12: 122801646-122801646 on Build GRCh38
rs770229576 Chr.12: 122801592-122801598 on Build GRCh38
rs201037164 Chr.12: 122801607-122801607 on Build GRCh38
rs780840811 Chr.12: 122801460-122801460 on Build GRCh38
rs748269523 Chr.12: 122801516-122801516 on Build GRCh38
rs778139001 Chr.12: 122801636-122801636 on Build GRCh38
rs368745484 Chr.12: 122801561-122801561 on Build GRCh38
rs150261986 Chr.12: 122801478-122801478 on Build GRCh38
rs747294737 Chr.12: 122801639-122801639 on Build GRCh38
rs199705995 Chr.12: 122801465-122801465 on Build GRCh38
rs1034934949 Chr.12: 122801475-122801475 on Build GRCh38
rs759979825 Chr.12: 122801659-122801659 on Build GRCh38
rs754830884 Chr.12: 122801612-122801612 on Build GRCh38
rs985198794 Chr.12: 122801518-122801518 on Build GRCh38

View Full Product Details