Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138946076-138946313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXL2
Entrez Gene ID
668
Gene Name
forkhead box L2
Gene Aliases
BPES, BPES1, PFRK, PINTO, POF3
UniGene
Hs.289292
Gene Chromosome Location
Chr.3: 138944224-138947140 on Build GRCh38
Transcripts
NM_023067.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6923817 c.520G>A p.G174S Chr.3: 138946203-138946203 on Build GRCh38
5449792 c.520G>T p.G174C Chr.3: 138946203-138946203 on Build GRCh38
4600643 c.536C>G p.A179G Chr.3: 138946187-138946187 on Build GRCh38
6923876 c.434G>T p.R145L Chr.3: 138946289-138946289 on Build GRCh38
1419458 c.433C>T p.R145C Chr.3: 138946290-138946290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894046852 Chr.3: 138946197-138946197 on Build GRCh38
rs779956639 Chr.3: 138946114-138946114 on Build GRCh38
rs1057516162 Chr.3: 138946145-138946145 on Build GRCh38
rs758494065 Chr.3: 138946109-138946109 on Build GRCh38
rs747079191 Chr.3: 138946157-138946157 on Build GRCh38
rs770228060 Chr.3: 138946210-138946210 on Build GRCh38
rs1057516164 Chr.3: 138946111-138946111 on Build GRCh38
rs121908359 Chr.3: 138946163-138946163 on Build GRCh38
rs781285704 Chr.3: 138946285-138946285 on Build GRCh38
rs761606348 Chr.3: 138946237-138946237 on Build GRCh38
rs1057516161 Chr.3: 138946146-138946147 on Build GRCh38
rs1029944805 Chr.3: 138946288-138946288 on Build GRCh38
rs760223092 Chr.3: 138946227-138946227 on Build GRCh38
rs1057516165 Chr.3: 138946105-138946105 on Build GRCh38
rs774916125 Chr.3: 138946225-138946225 on Build GRCh38
rs955713442 Chr.3: 138946293-138946293 on Build GRCh38
rs750388967 Chr.3: 138946252-138946252 on Build GRCh38
rs61750361 Chr.3: 138946222-138946222 on Build GRCh38
rs776277339 Chr.3: 138946236-138946236 on Build GRCh38
rs768808919 Chr.3: 138946159-138946159 on Build GRCh38
rs1057516163 Chr.3: 138946141-138946141 on Build GRCh38
rs928445396 Chr.3: 138946189-138946189 on Build GRCh38
rs560949046 Chr.3: 138946201-138946201 on Build GRCh38
rs755018932 Chr.3: 138946278-138946278 on Build GRCh38
rs915586947 Chr.3: 138946162-138946162 on Build GRCh38
rs749775902 Chr.3: 138946221-138946221 on Build GRCh38
rs763716595 Chr.3: 138946228-138946228 on Build GRCh38
rs104893739 Chr.3: 138946137-138946137 on Build GRCh38
rs1019605023 Chr.3: 138946250-138946250 on Build GRCh38
rs997480684 Chr.3: 138946279-138946279 on Build GRCh38
rs766317905 Chr.3: 138946258-138946258 on Build GRCh38
rs954048795 Chr.3: 138946207-138946207 on Build GRCh38
rs929179236 Chr.3: 138946198-138946198 on Build GRCh38
rs751648653 Chr.3: 138946261-138946261 on Build GRCh38
rs572394953 Chr.3: 138946180-138946180 on Build GRCh38
rs7432551 Chr.3: 138946187-138946187 on Build GRCh38
rs758370933 Chr.3: 138946254-138946254 on Build GRCh38
rs529612732 Chr.3: 138946218-138946218 on Build GRCh38
rs957033721 Chr.3: 138946148-138946148 on Build GRCh38
rs764960739 Chr.3: 138946238-138946238 on Build GRCh38

View Full Product Details