Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92134045-92134306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF2
Entrez Gene ID
53981
Gene Name
cleavage and polyadenylation specific factor 2
Gene Aliases
CPSF100
UniGene
Hs.657632
Gene Chromosome Location
Chr.14: 92121954-92164199 on Build GRCh38
Transcripts
NM_001322270.1 NM_001322271.1 NM_001322272.1 NM_017437.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7288814 c.309+2T>C p.? Chr.14: 92134172-92134172 on Build GRCh38
4578064 c.365C>A p.A122D Chr.14: 92134305-92134305 on Build GRCh38
4954441 c.199C>A p.H67N Chr.14: 92134060-92134060 on Build GRCh38
5770847 c.184C>G p.H62D Chr.14: 92134045-92134045 on Build GRCh38
6634844 c.225C>T p.V75V Chr.14: 92134086-92134086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200651259 Chr.14: 92134276-92134276 on Build GRCh38
rs745957916 Chr.14: 92134260-92134260 on Build GRCh38
rs142923243 Chr.14: 92134160-92134160 on Build GRCh38
rs748515128 Chr.14: 92134153-92134153 on Build GRCh38
rs984701274 Chr.14: 92134087-92134087 on Build GRCh38
rs373232696 Chr.14: 92134219-92134219 on Build GRCh38
rs372145276 Chr.14: 92134076-92134076 on Build GRCh38
rs890061655 Chr.14: 92134277-92134277 on Build GRCh38
rs761608291 Chr.14: 92134193-92134193 on Build GRCh38
rs756029510 Chr.14: 92134207-92134207 on Build GRCh38
rs747456628 Chr.14: 92134165-92134165 on Build GRCh38
rs769969271 Chr.14: 92134279-92134279 on Build GRCh38
rs182853150 Chr.14: 92134241-92134241 on Build GRCh38
rs141630712 Chr.14: 92134077-92134077 on Build GRCh38
rs138644568 Chr.14: 92134113-92134113 on Build GRCh38
rs769714806 Chr.14: 92134228-92134231 on Build GRCh38
rs150522138 Chr.14: 92134086-92134086 on Build GRCh38
rs755623894 Chr.14: 92134097-92134097 on Build GRCh38
rs767230801 Chr.14: 92134194-92134194 on Build GRCh38
rs201289801 Chr.14: 92134202-92134202 on Build GRCh38
rs1056451968 Chr.14: 92134152-92134152 on Build GRCh38
rs772767378 Chr.14: 92134155-92134155 on Build GRCh38
rs1004327102 Chr.14: 92134171-92134171 on Build GRCh38
rs778247450 Chr.14: 92134250-92134250 on Build GRCh38
rs887302335 Chr.14: 92134154-92134154 on Build GRCh38
rs771448049 Chr.14: 92134255-92134255 on Build GRCh38
rs116318468 Chr.14: 92134212-92134212 on Build GRCh38
rs768328034 Chr.14: 92134120-92134120 on Build GRCh38
rs776725548 Chr.14: 92134180-92134180 on Build GRCh38
rs1017174135 Chr.14: 92134195-92134195 on Build GRCh38
rs748038439 Chr.14: 92134186-92134188 on Build GRCh38
rs763819164 Chr.14: 92134068-92134068 on Build GRCh38
rs376631823 Chr.14: 92134176-92134176 on Build GRCh38
rs747545780 Chr.14: 92134252-92134252 on Build GRCh38
rs373374534 Chr.14: 92134182-92134182 on Build GRCh38
rs776614411 Chr.14: 92134258-92134258 on Build GRCh38
rs779183068 Chr.14: 92134123-92134123 on Build GRCh38
rs779575721 Chr.14: 92134099-92134099 on Build GRCh38
rs765469633 Chr.14: 92134191-92134191 on Build GRCh38
rs774200294 Chr.14: 92134192-92134192 on Build GRCh38
rs376143459 Chr.14: 92134229-92134229 on Build GRCh38
rs868609987 Chr.14: 92134177-92134177 on Build GRCh38
rs753354408 Chr.14: 92134218-92134218 on Build GRCh38

View Full Product Details