Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17119817-17120046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR6
Entrez Gene ID
27439
Gene Name
cat eye syndrome chromosome region, candidate 6
Gene Aliases
-
UniGene
Hs.209577
Gene Chromosome Location
Chr.22: 17116299-17121367 on Build GRCh38
Transcripts
NM_001163079.1 NM_031890.3 XM_011546124.2 XM_011546125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4102364 c.1123G>A p.A375T Chr.22: 17120005-17120005 on Build GRCh38
5584769 c.1161C>T p.P387P Chr.22: 17119967-17119967 on Build GRCh38
4102363 c.1165C>T p.R389W Chr.22: 17119963-17119963 on Build GRCh38
5555401 c.1120G>A p.G374S Chr.22: 17120008-17120008 on Build GRCh38
4639497 c.1233G>A p.E411E Chr.22: 17119895-17119895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747913031 Chr.22: 17119858-17119858 on Build GRCh38
rs752161859 Chr.22: 17119931-17119931 on Build GRCh38
rs764683551 Chr.22: 17119910-17119910 on Build GRCh38
rs754095956 Chr.22: 17120026-17120026 on Build GRCh38
rs369034149 Chr.22: 17119953-17119953 on Build GRCh38
rs777652229 Chr.22: 17119975-17119975 on Build GRCh38
rs746142298 Chr.22: 17119849-17119849 on Build GRCh38
rs756304254 Chr.22: 17119948-17119948 on Build GRCh38
rs372152443 Chr.22: 17119865-17119865 on Build GRCh38
rs931193134 Chr.22: 17119864-17119864 on Build GRCh38
rs767666662 Chr.22: 17119933-17119933 on Build GRCh38
rs373040255 Chr.22: 17119968-17119968 on Build GRCh38
rs374898503 Chr.22: 17119951-17119951 on Build GRCh38
rs776032998 Chr.22: 17119878-17119878 on Build GRCh38
rs376508104 Chr.22: 17119888-17119888 on Build GRCh38
rs560609218 Chr.22: 17120002-17120002 on Build GRCh38
rs377076603 Chr.22: 17119984-17119984 on Build GRCh38
rs755219751 Chr.22: 17120027-17120027 on Build GRCh38
rs368814541 Chr.22: 17119838-17119838 on Build GRCh38
rs776148578 Chr.22: 17119991-17119991 on Build GRCh38
rs770463927 Chr.22: 17119874-17119874 on Build GRCh38
rs530348909 Chr.22: 17119997-17119997 on Build GRCh38
rs769940818 Chr.22: 17119851-17119851 on Build GRCh38
rs778724970 Chr.22: 17119854-17119854 on Build GRCh38
rs745373554 Chr.22: 17119995-17119995 on Build GRCh38
rs998981423 Chr.22: 17119943-17119943 on Build GRCh38
rs893143980 Chr.22: 17119952-17119952 on Build GRCh38
rs901919965 Chr.22: 17119939-17119939 on Build GRCh38
rs750492093 Chr.22: 17119944-17119944 on Build GRCh38
rs756218826 Chr.22: 17120017-17120017 on Build GRCh38
rs140508260 Chr.22: 17119871-17119871 on Build GRCh38
rs750453167 Chr.22: 17120007-17120007 on Build GRCh38
rs760152806 Chr.22: 17119873-17119873 on Build GRCh38
rs970768108 Chr.22: 17119980-17119980 on Build GRCh38
rs757381613 Chr.22: 17119932-17119932 on Build GRCh38
rs759176352 Chr.22: 17119880-17119880 on Build GRCh38
rs369425031 Chr.22: 17119895-17119895 on Build GRCh38
rs202143826 Chr.22: 17120000-17120000 on Build GRCh38
rs552494738 Chr.22: 17120018-17120018 on Build GRCh38
rs1040909265 Chr.22: 17119935-17119935 on Build GRCh38
rs763477547 Chr.22: 17119901-17119901 on Build GRCh38
rs371295778 Chr.22: 17120005-17120005 on Build GRCh38
rs371691472 Chr.22: 17119937-17119937 on Build GRCh38
rs763389011 Chr.22: 17119999-17119999 on Build GRCh38

View Full Product Details