Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62702363-62702618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSCL2
Entrez Gene ID
26580
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Gene Aliases
GNG3LG, HMN5, PELD, SPG17
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62709619 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613135 c.260C>T p.A87V Chr.11: 62702502-62702502 on Build GRCh38
6866332 c.279G>T p.K93N Chr.11: 62702483-62702483 on Build GRCh38
4546871 c.217G>A p.D73N Chr.11: 62702545-62702545 on Build GRCh38
4546870 c.409G>A p.D137N Chr.11: 62702545-62702545 on Build GRCh38
6613134 c.452C>T p.A151V Chr.11: 62702502-62702502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533688134 Chr.11: 62702573-62702573 on Build GRCh38
rs763484529 Chr.11: 62702507-62702507 on Build GRCh38
rs748460314 Chr.11: 62702566-62702566 on Build GRCh38
rs778111224 Chr.11: 62702569-62702569 on Build GRCh38
rs137852973 Chr.11: 62702493-62702493 on Build GRCh38
rs137930278 Chr.11: 62702488-62702488 on Build GRCh38
rs777165971 Chr.11: 62702575-62702575 on Build GRCh38
rs137852972 Chr.11: 62702499-62702499 on Build GRCh38
rs751277966 Chr.11: 62702511-62702511 on Build GRCh38
rs761979025 Chr.11: 62702409-62702409 on Build GRCh38
rs773733821 Chr.11: 62702588-62702588 on Build GRCh38
rs780093151 Chr.11: 62702534-62702534 on Build GRCh38
rs749324787 Chr.11: 62702459-62702459 on Build GRCh38
rs766801709 Chr.11: 62702492-62702492 on Build GRCh38
rs755125013 Chr.11: 62702555-62702555 on Build GRCh38
rs893647812 Chr.11: 62702466-62702466 on Build GRCh38
rs767275692 Chr.11: 62702432-62702432 on Build GRCh38
rs761566669 Chr.11: 62702575-62702577 on Build GRCh38
rs778835265 Chr.11: 62702561-62702561 on Build GRCh38
rs879253900 Chr.11: 62702545-62702545 on Build GRCh38
rs768556778 Chr.11: 62702562-62702564 on Build GRCh38
rs747313556 Chr.11: 62702572-62702572 on Build GRCh38
rs774015823 Chr.11: 62702565-62702565 on Build GRCh38
rs187108650 Chr.11: 62702582-62702582 on Build GRCh38
rs768780378 Chr.11: 62702469-62702469 on Build GRCh38
rs780831457 Chr.11: 62702430-62702430 on Build GRCh38
rs748287282 Chr.11: 62702475-62702475 on Build GRCh38
rs774590929 Chr.11: 62702470-62702470 on Build GRCh38
rs754001369 Chr.11: 62702546-62702546 on Build GRCh38
rs72929434 Chr.11: 62702457-62702457 on Build GRCh38
rs1017482651 Chr.11: 62702563-62702563 on Build GRCh38
rs144774892 Chr.11: 62702522-62702522 on Build GRCh38
rs149412531 Chr.11: 62702506-62702506 on Build GRCh38
rs1003234218 Chr.11: 62702502-62702502 on Build GRCh38
rs772536764 Chr.11: 62702476-62702476 on Build GRCh38
rs750176844 Chr.11: 62702529-62702529 on Build GRCh38
rs773548699 Chr.11: 62702485-62702485 on Build GRCh38
rs1011200048 Chr.11: 62702467-62702467 on Build GRCh38
rs973365158 Chr.11: 62702577-62702577 on Build GRCh38
rs267603078 Chr.11: 62702513-62702513 on Build GRCh38
rs762779983 Chr.11: 62702518-62702519 on Build GRCh38
rs756117331 Chr.11: 62702439-62702439 on Build GRCh38
rs1020744035 Chr.11: 62702428-62702428 on Build GRCh38
rs756135904 Chr.11: 62702531-62702531 on Build GRCh38
rs767486914 Chr.11: 62702526-62702526 on Build GRCh38

View Full Product Details