Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132078124-132078397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
UniGene
Hs.170776
Gene Chromosome Location
Chr.X: 132076986-132128022 on Build GRCh38
Transcripts
NM_001306193.1 NM_194277.2 XM_017029947.1 XM_017029948.1 XM_017029949.1

SNP Detail

SNP ID Chromosome Location
rs779971174 Chr.X: 132078219-132078219 on Build GRCh38
rs754559834 Chr.X: 132078328-132078328 on Build GRCh38
rs764829601 Chr.X: 132078266-132078266 on Build GRCh38
rs760487000 Chr.X: 132078250-132078250 on Build GRCh38
rs951558941 Chr.X: 132078233-132078233 on Build GRCh38
rs751858239 Chr.X: 132078146-132078146 on Build GRCh38
rs369003419 Chr.X: 132078343-132078343 on Build GRCh38
rs746083026 Chr.X: 132078212-132078212 on Build GRCh38
rs746914277 Chr.X: 132078361-132078361 on Build GRCh38
rs980956013 Chr.X: 132078356-132078356 on Build GRCh38
rs751994722 Chr.X: 132078218-132078218 on Build GRCh38
rs781502273 Chr.X: 132078302-132078302 on Build GRCh38
rs770900557 Chr.X: 132078373-132078373 on Build GRCh38
rs757487294 Chr.X: 132078185-132078185 on Build GRCh38
rs868852708 Chr.X: 132078213-132078213 on Build GRCh38
rs1029569386 Chr.X: 132078282-132078282 on Build GRCh38
rs748139262 Chr.X: 132078230-132078230 on Build GRCh38
rs772057382 Chr.X: 132078353-132078353 on Build GRCh38
rs776464042 Chr.X: 132078255-132078255 on Build GRCh38
rs1025726284 Chr.X: 132078225-132078225 on Build GRCh38
rs752045112 Chr.X: 132078287-132078287 on Build GRCh38
rs962501934 Chr.X: 132078349-132078349 on Build GRCh38
rs773063259 Chr.X: 132078240-132078240 on Build GRCh38
rs781449121 Chr.X: 132078201-132078201 on Build GRCh38
rs776519156 Chr.X: 132078374-132078374 on Build GRCh38
rs756241221 Chr.X: 132078325-132078325 on Build GRCh38
rs903939499 Chr.X: 132078203-132078203 on Build GRCh38
rs771864320 Chr.X: 132078238-132078238 on Build GRCh38
rs759241920 Chr.X: 132078259-132078259 on Build GRCh38
rs750688292 Chr.X: 132078312-132078312 on Build GRCh38
rs140383991 Chr.X: 132078342-132078342 on Build GRCh38
rs1039105692 Chr.X: 132078223-132078223 on Build GRCh38
rs375348707 Chr.X: 132078152-132078152 on Build GRCh38
rs777640376 Chr.X: 132078357-132078357 on Build GRCh38
rs770847430 Chr.X: 132078251-132078251 on Build GRCh38
rs757682240 Chr.X: 132078300-132078300 on Build GRCh38
rs148279797 Chr.X: 132078276-132078276 on Build GRCh38
rs748187461 Chr.X: 132078213-132078218 on Build GRCh38
rs763523180 Chr.X: 132078273-132078273 on Build GRCh38
rs1003052816 Chr.X: 132078258-132078258 on Build GRCh38
rs1043882590 Chr.X: 132078168-132078168 on Build GRCh38
rs752113123 Chr.X: 132078267-132078267 on Build GRCh38

View Full Product Details