Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41917085-41917358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf51
Entrez Gene ID
285636
Gene Name
chromosome 5 open reading frame 51
Gene Aliases
-
UniGene
Hs.29899
Gene Chromosome Location
Chr.5: 41904313-41921636 on Build GRCh38
Transcripts
NM_175921.5 XM_005248289.4 XM_011514032.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3342386 c.727G>T p.G243C Chr.5: 41917141-41917141 on Build GRCh38
3342387 c.856T>G p.L286V Chr.5: 41917270-41917270 on Build GRCh38
1068336 c.808G>T p.G270* Chr.5: 41917222-41917222 on Build GRCh38
6616342 c.799G>C p.V267L Chr.5: 41917213-41917213 on Build GRCh38
6616343 c.672T>C p.Y224Y Chr.5: 41917086-41917086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529612474 Chr.5: 41917182-41917182 on Build GRCh38
rs763195986 Chr.5: 41917186-41917186 on Build GRCh38
rs149416451 Chr.5: 41917203-41917203 on Build GRCh38
rs772461387 Chr.5: 41917296-41917308 on Build GRCh38
rs776627931 Chr.5: 41917174-41917174 on Build GRCh38
rs747172856 Chr.5: 41917147-41917147 on Build GRCh38
rs764727957 Chr.5: 41917191-41917191 on Build GRCh38
rs769954153 Chr.5: 41917176-41917176 on Build GRCh38
rs754729263 Chr.5: 41917218-41917218 on Build GRCh38
rs965867008 Chr.5: 41917167-41917167 on Build GRCh38
rs762319396 Chr.5: 41917210-41917210 on Build GRCh38
rs34949951 Chr.5: 41917198-41917198 on Build GRCh38
rs757148532 Chr.5: 41917309-41917309 on Build GRCh38
rs779908571 Chr.5: 41917113-41917113 on Build GRCh38
rs978823453 Chr.5: 41917253-41917253 on Build GRCh38
rs774703684 Chr.5: 41917199-41917199 on Build GRCh38
rs61731719 Chr.5: 41917140-41917140 on Build GRCh38
rs771011863 Chr.5: 41917151-41917151 on Build GRCh38
rs368328564 Chr.5: 41917212-41917212 on Build GRCh38
rs753549498 Chr.5: 41917122-41917122 on Build GRCh38
rs1057375494 Chr.5: 41917289-41917289 on Build GRCh38
rs759786958 Chr.5: 41917175-41917175 on Build GRCh38
rs138879731 Chr.5: 41917143-41917143 on Build GRCh38
rs745924482 Chr.5: 41917322-41917322 on Build GRCh38
rs912579402 Chr.5: 41917120-41917120 on Build GRCh38
rs758118598 Chr.5: 41917275-41917275 on Build GRCh38
rs752608448 Chr.5: 41917254-41917254 on Build GRCh38
rs753355135 Chr.5: 41917215-41917215 on Build GRCh38
rs199832727 Chr.5: 41917145-41917145 on Build GRCh38
rs746213142 Chr.5: 41917160-41917160 on Build GRCh38
rs770042531 Chr.5: 41917324-41917324 on Build GRCh38
rs144285732 Chr.5: 41917288-41917288 on Build GRCh38
rs1041923386 Chr.5: 41917211-41917211 on Build GRCh38
rs754606695 Chr.5: 41917139-41917139 on Build GRCh38
rs374722484 Chr.5: 41917320-41917320 on Build GRCh38
rs746926407 Chr.5: 41917306-41917306 on Build GRCh38
rs777570944 Chr.5: 41917146-41917146 on Build GRCh38
rs758682717 Chr.5: 41917142-41917142 on Build GRCh38
rs368512609 Chr.5: 41917257-41917257 on Build GRCh38
rs376745817 Chr.5: 41917239-41917239 on Build GRCh38

View Full Product Details