Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78221590-78221851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB2
Entrez Gene ID
9846
Gene Name
GRB2 associated binding protein 2
Gene Aliases
-
UniGene
Hs.429434
Gene Chromosome Location
Chr.11: 78215290-78417822 on Build GRCh38
Transcripts
NM_012296.3 NM_080491.2 XM_006718753.2 XM_011545408.2 XM_017018052.1

Gene Symbol
USP35
Entrez Gene ID
57558
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
-
UniGene
Hs.531249
Gene Chromosome Location
Chr.11: 78188812-78233651 on Build GRCh38
Transcripts
NM_012296.3 NM_080491.2 XM_006718753.2 XM_011545408.2 XM_017018052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5373661 c.1726G>A p.D576N Chr.11: 78221712-78221712 on Build GRCh38
4530862 c.1735G>A p.D579N Chr.11: 78221703-78221703 on Build GRCh38
932277 c.1693C>T p.R565C Chr.11: 78221745-78221745 on Build GRCh38
4790333 c.1712G>T p.S571I Chr.11: 78221726-78221726 on Build GRCh38
5027451 c.1735G>C p.D579H Chr.11: 78221703-78221703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937683104 Chr.11: 78221623-78221623 on Build GRCh38
rs141147842 Chr.11: 78221827-78221827 on Build GRCh38
rs760207545 Chr.11: 78221670-78221670 on Build GRCh38
rs764584872 Chr.11: 78221685-78221685 on Build GRCh38
rs114514318 Chr.11: 78221710-78221710 on Build GRCh38
rs757545082 Chr.11: 78221636-78221636 on Build GRCh38
rs889841923 Chr.11: 78221739-78221739 on Build GRCh38
rs778979203 Chr.11: 78221728-78221728 on Build GRCh38
rs767669827 Chr.11: 78221703-78221703 on Build GRCh38
rs563245503 Chr.11: 78221744-78221744 on Build GRCh38
rs367549795 Chr.11: 78221658-78221658 on Build GRCh38
rs994742841 Chr.11: 78221803-78221803 on Build GRCh38
rs771678554 Chr.11: 78221666-78221666 on Build GRCh38
rs541168313 Chr.11: 78221648-78221648 on Build GRCh38
rs149853536 Chr.11: 78221655-78221655 on Build GRCh38
rs755030706 Chr.11: 78221727-78221727 on Build GRCh38
rs983457877 Chr.11: 78221789-78221789 on Build GRCh38
rs747702687 Chr.11: 78221731-78221731 on Build GRCh38
rs1026180199 Chr.11: 78221821-78221821 on Build GRCh38
rs766575221 Chr.11: 78221787-78221787 on Build GRCh38
rs368907942 Chr.11: 78221692-78221692 on Build GRCh38
rs907902913 Chr.11: 78221817-78221817 on Build GRCh38
rs476687 Chr.11: 78221755-78221755 on Build GRCh38
rs548988387 Chr.11: 78221795-78221795 on Build GRCh38
rs766362187 Chr.11: 78221735-78221735 on Build GRCh38
rs767655048 Chr.11: 78221823-78221823 on Build GRCh38
rs1006906902 Chr.11: 78221757-78221757 on Build GRCh38
rs746689935 Chr.11: 78221748-78221748 on Build GRCh38
rs764637960 Chr.11: 78221790-78221790 on Build GRCh38
rs500942 Chr.11: 78221793-78221793 on Build GRCh38
rs765656562 Chr.11: 78221674-78221674 on Build GRCh38
rs200015699 Chr.11: 78221747-78221747 on Build GRCh38
rs755419738 Chr.11: 78221665-78221665 on Build GRCh38
rs147686701 Chr.11: 78221679-78221679 on Build GRCh38
rs2848984 Chr.11: 78221681-78221681 on Build GRCh38
rs750684716 Chr.11: 78221705-78221705 on Build GRCh38
rs946953372 Chr.11: 78221662-78221662 on Build GRCh38
rs749023909 Chr.11: 78221722-78221722 on Build GRCh38
rs779670406 Chr.11: 78221652-78221652 on Build GRCh38
rs781465416 Chr.11: 78221639-78221639 on Build GRCh38
rs145840199 Chr.11: 78221782-78221782 on Build GRCh38
rs1809532 Chr.11: 78221633-78221633 on Build GRCh38
rs762183376 Chr.11: 78221816-78221816 on Build GRCh38
rs142438889 Chr.11: 78221772-78221772 on Build GRCh38
rs376071505 Chr.11: 78221712-78221712 on Build GRCh38
rs772890796 Chr.11: 78221667-78221667 on Build GRCh38
rs774002523 Chr.11: 78221659-78221659 on Build GRCh38
rs757341769 Chr.11: 78221698-78221698 on Build GRCh38
rs745696170 Chr.11: 78221644-78221644 on Build GRCh38
rs76023882 Chr.11: 78221783-78221783 on Build GRCh38
rs141039033 Chr.11: 78221618-78221618 on Build GRCh38

View Full Product Details