Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168085592-168085837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR161
Entrez Gene ID
23432
Gene Name
G protein-coupled receptor 161
Gene Aliases
RE2
UniGene
Hs.271809
Gene Chromosome Location
Chr.1: 168079542-168137667 on Build GRCh38
Transcripts
NM_001267609.1 NM_001267610.1 NM_001267611.1 NM_001267612.1 NM_001267613.1 NM_001267614.1 NM_153832.2 XM_005245055.2 XM_005245056.2 XM_005245057.4 XM_006711251.1 XM_006711253.2 XM_011509370.1 XM_011509371.2 XM_011509372.1 XM_011509373.2 XM_011509375.2 XM_011509376.2 XM_011509377.1 XM_011509378.1 XM_017000858.1 XM_017000859.1 XM_017000860.1 XM_017000861.1 XM_017000862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7114593 c.1086C>T p.I362I Chr.1: 168085693-168085693 on Build GRCh38
7264257 c.1264G>A p.G422R Chr.1: 168085623-168085623 on Build GRCh38
4167656 c.1490G>A p.G497D Chr.1: 168085631-168085631 on Build GRCh38
7114590 c.1488C>T p.I496I Chr.1: 168085693-168085693 on Build GRCh38
4607417 c.983-1G>T p.? Chr.1: 168085797-168085797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77478968 Chr.1: 168085813-168085813 on Build GRCh38
rs200521694 Chr.1: 168085652-168085652 on Build GRCh38
rs767869653 Chr.1: 168085753-168085753 on Build GRCh38
rs752799929 Chr.1: 168085758-168085758 on Build GRCh38
rs759927031 Chr.1: 168085752-168085752 on Build GRCh38
rs148848526 Chr.1: 168085616-168085616 on Build GRCh38
rs779748525 Chr.1: 168085803-168085803 on Build GRCh38
rs760237330 Chr.1: 168085715-168085715 on Build GRCh38
rs373775972 Chr.1: 168085630-168085630 on Build GRCh38
rs746627581 Chr.1: 168085807-168085807 on Build GRCh38
rs897155227 Chr.1: 168085618-168085618 on Build GRCh38
rs774108367 Chr.1: 168085705-168085705 on Build GRCh38
rs766670512 Chr.1: 168085619-168085619 on Build GRCh38
rs765787301 Chr.1: 168085639-168085639 on Build GRCh38
rs373399666 Chr.1: 168085676-168085676 on Build GRCh38
rs146091627 Chr.1: 168085637-168085637 on Build GRCh38
rs145492270 Chr.1: 168085614-168085614 on Build GRCh38
rs930194782 Chr.1: 168085623-168085623 on Build GRCh38
rs199866265 Chr.1: 168085704-168085704 on Build GRCh38
rs753864637 Chr.1: 168085779-168085779 on Build GRCh38
rs759173797 Chr.1: 168085632-168085632 on Build GRCh38
rs142783286 Chr.1: 168085647-168085647 on Build GRCh38
rs894924999 Chr.1: 168085686-168085686 on Build GRCh38
rs751881848 Chr.1: 168085654-168085654 on Build GRCh38
rs535791878 Chr.1: 168085638-168085638 on Build GRCh38
rs755719942 Chr.1: 168085735-168085735 on Build GRCh38
rs745558794 Chr.1: 168085707-168085707 on Build GRCh38
rs573642610 Chr.1: 168085771-168085771 on Build GRCh38
rs1000792342 Chr.1: 168085772-168085772 on Build GRCh38
rs766887809 Chr.1: 168085735-168085735 on Build GRCh38
rs918973203 Chr.1: 168085671-168085671 on Build GRCh38
rs752000379 Chr.1: 168085748-168085748 on Build GRCh38
rs755222954 Chr.1: 168085655-168085655 on Build GRCh38
rs763343811 Chr.1: 168085732-168085732 on Build GRCh38
rs1001678771 Chr.1: 168085646-168085646 on Build GRCh38
rs147976800 Chr.1: 168085631-168085631 on Build GRCh38
rs1022341428 Chr.1: 168085706-168085706 on Build GRCh38
rs745685551 Chr.1: 168085796-168085796 on Build GRCh38
rs367790399 Chr.1: 168085725-168085725 on Build GRCh38
rs148708325 Chr.1: 168085711-168085711 on Build GRCh38
rs184385549 Chr.1: 168085780-168085780 on Build GRCh38
rs201690810 Chr.1: 168085712-168085712 on Build GRCh38
rs748265409 Chr.1: 168085675-168085675 on Build GRCh38
rs528609467 Chr.1: 168085624-168085624 on Build GRCh38
rs112752784 Chr.1: 168085708-168085708 on Build GRCh38
rs781528140 Chr.1: 168085672-168085672 on Build GRCh38
rs376463579 Chr.1: 168085781-168085781 on Build GRCh38
rs758866074 Chr.1: 168085649-168085649 on Build GRCh38
rs767061233 Chr.1: 168085636-168085636 on Build GRCh38
rs748140638 Chr.1: 168085613-168085613 on Build GRCh38
rs763509138 Chr.1: 168085717-168085717 on Build GRCh38
rs1014328026 Chr.1: 168085719-168085719 on Build GRCh38
rs763533709 Chr.1: 168085812-168085813 on Build GRCh38
rs749174723 Chr.1: 168085700-168085700 on Build GRCh38
rs777767557 Chr.1: 168085685-168085685 on Build GRCh38
rs752347222 Chr.1: 168085630-168085631 on Build GRCh38
rs777703825 Chr.1: 168085777-168085777 on Build GRCh38
rs1010570425 Chr.1: 168085702-168085702 on Build GRCh38
rs771788199 Chr.1: 168085801-168085801 on Build GRCh38

View Full Product Details