Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104880164-104880431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170B
Entrez Gene ID
283638
Gene Name
centrosomal protein 170B
Gene Aliases
CEP170R, FAM68C, KIAA0284
UniGene
Hs.533721
Gene Chromosome Location
Chr.14: 104865313-104896770 on Build GRCh38
Transcripts
NM_001112726.2 NM_015005.2 XM_005267550.3 XM_011536665.2 XM_011536666.2 XM_017021227.1 XM_017021228.1 XM_017021229.1 XM_017021230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420292 c.455A>G p.D152G Chr.14: 104880408-104880408 on Build GRCh38
6396608 c.262G>C p.E88Q Chr.14: 104880425-104880425 on Build GRCh38
3711463 c.124-1G>T p.? Chr.14: 104880286-104880286 on Build GRCh38
3711462 c.334-1G>T p.? Chr.14: 104880286-104880286 on Build GRCh38
4405620 c.228C>T p.P76P Chr.14: 104880391-104880391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986172328 Chr.14: 104880382-104880382 on Build GRCh38
rs868205651 Chr.14: 104880310-104880310 on Build GRCh38
rs199634547 Chr.14: 104880333-104880333 on Build GRCh38
rs765432805 Chr.14: 104880410-104880410 on Build GRCh38
rs767591976 Chr.14: 104880266-104880267 on Build GRCh38
rs778194664 Chr.14: 104880364-104880364 on Build GRCh38
rs114271411 Chr.14: 104880270-104880270 on Build GRCh38
rs776008979 Chr.14: 104880319-104880319 on Build GRCh38
rs377138069 Chr.14: 104880262-104880262 on Build GRCh38
rs758489866 Chr.14: 104880412-104880412 on Build GRCh38
rs755278543 Chr.14: 104880346-104880346 on Build GRCh38
rs190671864 Chr.14: 104880396-104880396 on Build GRCh38
rs754083457 Chr.14: 104880339-104880339 on Build GRCh38
rs888517717 Chr.14: 104880269-104880269 on Build GRCh38
rs762471036 Chr.14: 104880384-104880384 on Build GRCh38
rs373618030 Chr.14: 104880377-104880377 on Build GRCh38
rs777018790 Chr.14: 104880375-104880375 on Build GRCh38
rs554588587 Chr.14: 104880266-104880266 on Build GRCh38
rs574900996 Chr.14: 104880242-104880242 on Build GRCh38
rs768383267 Chr.14: 104880335-104880335 on Build GRCh38
rs759297564 Chr.14: 104880308-104880308 on Build GRCh38
rs767399120 Chr.14: 104880394-104880394 on Build GRCh38
rs376305196 Chr.14: 104880358-104880358 on Build GRCh38
rs775746187 Chr.14: 104880265-104880266 on Build GRCh38
rs747571621 Chr.14: 104880367-104880367 on Build GRCh38
rs558430615 Chr.14: 104880225-104880225 on Build GRCh38
rs911441631 Chr.14: 104880192-104880192 on Build GRCh38
rs771279385 Chr.14: 104880370-104880370 on Build GRCh38
rs929988178 Chr.14: 104880402-104880402 on Build GRCh38
rs370341304 Chr.14: 104880376-104880376 on Build GRCh38
rs1033162099 Chr.14: 104880265-104880265 on Build GRCh38
rs764049041 Chr.14: 104880320-104880320 on Build GRCh38
rs374526542 Chr.14: 104880357-104880357 on Build GRCh38
rs72700176 Chr.14: 104880295-104880295 on Build GRCh38
rs922693333 Chr.14: 104880231-104880231 on Build GRCh38
rs888436862 Chr.14: 104880263-104880263 on Build GRCh38
rs368059834 Chr.14: 104880385-104880385 on Build GRCh38
rs748581988 Chr.14: 104880267-104880267 on Build GRCh38
rs772582155 Chr.14: 104880268-104880268 on Build GRCh38
rs977292347 Chr.14: 104880226-104880226 on Build GRCh38
rs368280335 Chr.14: 104880397-104880397 on Build GRCh38
rs771297773 Chr.14: 104880284-104880284 on Build GRCh38
rs997846881 Chr.14: 104880301-104880301 on Build GRCh38
rs752297725 Chr.14: 104880309-104880309 on Build GRCh38
rs774428408 Chr.14: 104880391-104880391 on Build GRCh38
rs979018205 Chr.14: 104880188-104880188 on Build GRCh38
rs372939669 Chr.14: 104880411-104880411 on Build GRCh38
rs930060879 Chr.14: 104880232-104880232 on Build GRCh38
rs200306378 Chr.14: 104880347-104880347 on Build GRCh38
rs112026036 Chr.14: 104880392-104880392 on Build GRCh38
rs941333093 Chr.14: 104880264-104880264 on Build GRCh38
rs185370385 Chr.14: 104880334-104880334 on Build GRCh38

View Full Product Details