The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002592.2 | NP_002583.1 | 2 - 3 | 466 | 56 |
NM_182649.1 | NP_872590.1 | 1 - 2 | 430 | 56 | |
GenBank mRNA | AK300558.1 | - | 2 - 3 | 385 | 56 |
AK313286.1 | - | 1 - 2 | 387 | 56 | |
BC000491.2 | - | 1 - 2 | 372 | 56 | |
BC062439.1 | - | 1 - 2 | 387 | 56 | |
BG612192.1 | - | 1 - 2 | 432 | 56 | |
CR536501.1 | - | 1 - 2 | 227 | 56 | |
CR541799.1 | - | 1 - 2 | 227 | 56 | |
M15796.1 | - | 1 - 2 | 345 | 56 |