Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL8A2
Entrez Gene ID
1296
Gene Name
collagen type VIII alpha 2
Gene Aliases
FECD, FECD1, PPCD, PPCD2
Chromosome Location
Chr.1: 36095236 - 36126207 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005202.3 NP_005193.1 3 - 4 280 85
XM_005270477.3 XP_005270534.1 3 - 4 980 85
GenBank mRNA AK074129.1 - 1 - 2 198 85
BC096296.3 - 1 - 2 198 85

View Full Product Details