Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155937255-155937480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSO
Entrez Gene ID
1519
Gene Name
cathepsin O
Gene Aliases
CTSO1
UniGene
Hs.75262
Gene Chromosome Location
Chr.4: 155924118-155953896 on Build GRCh38
Transcripts
NM_001334.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1052551 c.580G>T p.E194* Chr.4: 155937456-155937456 on Build GRCh38
6637832 c.673A>G p.S225G Chr.4: 155937363-155937363 on Build GRCh38
6365386 c.559G>T p.V187L Chr.4: 155937477-155937477 on Build GRCh38
4123052 c.674G>T p.S225I Chr.4: 155937362-155937362 on Build GRCh38
1428208 c.618C>T p.Y206Y Chr.4: 155937418-155937418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756278867 Chr.4: 155937347-155937348 on Build GRCh38
rs376527696 Chr.4: 155937435-155937435 on Build GRCh38
rs780085147 Chr.4: 155937425-155937425 on Build GRCh38
rs769248102 Chr.4: 155937353-155937353 on Build GRCh38
rs763411023 Chr.4: 155937356-155937356 on Build GRCh38
rs150623992 Chr.4: 155937418-155937418 on Build GRCh38
rs148501038 Chr.4: 155937380-155937380 on Build GRCh38
rs373042303 Chr.4: 155937345-155937345 on Build GRCh38
rs772628160 Chr.4: 155937448-155937448 on Build GRCh38
rs752972814 Chr.4: 155937383-155937383 on Build GRCh38
rs754890474 Chr.4: 155937431-155937431 on Build GRCh38
rs777630281 Chr.4: 155937401-155937401 on Build GRCh38
rs373467360 Chr.4: 155937422-155937422 on Build GRCh38
rs751934098 Chr.4: 155937362-155937362 on Build GRCh38
rs199833785 Chr.4: 155937317-155937317 on Build GRCh38
rs961278758 Chr.4: 155937306-155937306 on Build GRCh38
rs769458051 Chr.4: 155937441-155937441 on Build GRCh38
rs79891510 Chr.4: 155937289-155937289 on Build GRCh38
rs766869927 Chr.4: 155937358-155937358 on Build GRCh38
rs139753097 Chr.4: 155937427-155937427 on Build GRCh38
rs946533544 Chr.4: 155937305-155937305 on Build GRCh38
rs772757813 Chr.4: 155937355-155937355 on Build GRCh38
rs112530305 Chr.4: 155937328-155937328 on Build GRCh38
rs200764150 Chr.4: 155937413-155937413 on Build GRCh38
rs777788781 Chr.4: 155937404-155937405 on Build GRCh38
rs748193984 Chr.4: 155937344-155937345 on Build GRCh38
rs141848549 Chr.4: 155937405-155937405 on Build GRCh38
rs376261835 Chr.4: 155937373-155937373 on Build GRCh38
rs915117986 Chr.4: 155937302-155937302 on Build GRCh38

View Full Product Details