The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017798.3 | NP_060268.2 | 4 - 5 | 1950 | 85 |
GenBank mRNA | AB055518.1 | - | 1 - 2 | 1291 | 85 |
AK000398.1 | - | 2 - 3 | 1111 | 85 | |
AK095795.1 | - | 4 - 5 | 1668 | 85 | |
AK300073.1 | - | 2 - 3 | 1680 | 85 | |
AL834366.1 | - | 1 - 2 | 1071 | 85 | |
BC003681.1 | - | 3 - 4 | 1624 | 85 | |
BC016920.2 | - | 1 - 2 | 1367 | 85 | |
BC025264.1 | - | 1 - 2 | 1487 | 85 | |
BC050284.1 | - | 4 - 5 | 1950 | 85 |