Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174308638-174308863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP44
Entrez Gene ID
80817
Gene Name
centrosomal protein 44
Gene Aliases
KIAA1712, PS1TP3
UniGene
Hs.555989
Gene Chromosome Location
Chr.4: 174283677-174334355 on Build GRCh38
Transcripts
NM_001040157.2 NM_001145314.1 XM_011532285.2 XM_011532286.2 XM_011532287.2 XM_017008654.1 XM_017008655.1 XM_017008656.1 XM_017008657.1 XM_017008658.1 XM_017008659.1 XM_017008660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3602072 c.653T>C p.V218A Chr.4: 174308834-174308834 on Build GRCh38
5528575 c.560C>T p.S187F Chr.4: 174308741-174308741 on Build GRCh38
6488213 c.565G>A p.D189N Chr.4: 174308746-174308746 on Build GRCh38
6400456 c.571T>A p.L191I Chr.4: 174308752-174308752 on Build GRCh38
3428346 c.526C>T p.R176C Chr.4: 174308707-174308707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975178994 Chr.4: 174308761-174308761 on Build GRCh38
rs147478051 Chr.4: 174308708-174308708 on Build GRCh38
rs763703431 Chr.4: 174308767-174308767 on Build GRCh38
rs528238232 Chr.4: 174308788-174308788 on Build GRCh38
rs761581759 Chr.4: 174308772-174308772 on Build GRCh38
rs760203338 Chr.4: 174308684-174308684 on Build GRCh38
rs760278137 Chr.4: 174308762-174308762 on Build GRCh38
rs766363239 Chr.4: 174308794-174308794 on Build GRCh38
rs370582883 Chr.4: 174308819-174308819 on Build GRCh38
rs757554027 Chr.4: 174308833-174308833 on Build GRCh38
rs751249478 Chr.4: 174308728-174308728 on Build GRCh38
rs139331696 Chr.4: 174308737-174308737 on Build GRCh38
rs778192446 Chr.4: 174308732-174308732 on Build GRCh38
rs373546998 Chr.4: 174308730-174308730 on Build GRCh38
rs377528697 Chr.4: 174308758-174308758 on Build GRCh38
rs746290439 Chr.4: 174308753-174308753 on Build GRCh38
rs148258730 Chr.4: 174308707-174308707 on Build GRCh38
rs774911680 Chr.4: 174308751-174308751 on Build GRCh38
rs73867207 Chr.4: 174308705-174308705 on Build GRCh38
rs756305464 Chr.4: 174308818-174308818 on Build GRCh38
rs766876653 Chr.4: 174308670-174308670 on Build GRCh38
rs764954886 Chr.4: 174308780-174308780 on Build GRCh38
rs752700084 Chr.4: 174308810-174308810 on Build GRCh38
rs757081528 Chr.4: 174308702-174308702 on Build GRCh38
rs370380836 Chr.4: 174308674-174308674 on Build GRCh38
rs775066766 Chr.4: 174308671-174308671 on Build GRCh38
rs561221477 Chr.4: 174308695-174308695 on Build GRCh38
rs771493509 Chr.4: 174308749-174308749 on Build GRCh38
rs375150474 Chr.4: 174308832-174308832 on Build GRCh38
rs75476714 Chr.4: 174308760-174308760 on Build GRCh38
rs765154254 Chr.4: 174308703-174308703 on Build GRCh38
rs184443103 Chr.4: 174308729-174308729 on Build GRCh38
rs776331014 Chr.4: 174308768-174308768 on Build GRCh38
rs750236678 Chr.4: 174308786-174308786 on Build GRCh38
rs753607384 Chr.4: 174308700-174308700 on Build GRCh38

View Full Product Details