The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176822.3 | NP_789792.1 | 10 - 11 | 3295 | 91 |
XM_011520044.1 | XP_011518346.1 | 9 - 10 | 3147 | 91 | |
GenBank mRNA | AK310900.1 | - | 10 - 11 | 3295 | 91 |
AY154469.1 | - | 9 - 10 | 2972 | 91 |