Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 41965540-41965781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI3
Entrez Gene ID
2737
Gene Name
GLI family zinc finger 3
Gene Aliases
ACLS, GCPS, GLI3-190, GLI3FL, PAP-A, PAPA, PAPA1, PAPB, PHS, PPDIV
UniGene
Hs.21509
Gene Chromosome Location
Chr.7: 41960949-42237019 on Build GRCh38
Transcripts
NM_000168.5 XM_011515274.2 XM_017011997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208298 c.3462C>A p.S1154R Chr.7: 41965611-41965611 on Build GRCh38
3431600 c.3396C>T p.P1132P Chr.7: 41965677-41965677 on Build GRCh38
1450715 c.3393G>A p.A1131A Chr.7: 41965680-41965680 on Build GRCh38
3229921 c.3444G>T p.Q1148H Chr.7: 41965629-41965629 on Build GRCh38
3431602 c.3312C>A p.N1104K Chr.7: 41965761-41965761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372503440 Chr.7: 41965622-41965622 on Build GRCh38
rs748255243 Chr.7: 41965699-41965699 on Build GRCh38
rs752183617 Chr.7: 41965578-41965578 on Build GRCh38
rs531098202 Chr.7: 41965723-41965723 on Build GRCh38
rs749508811 Chr.7: 41965620-41965620 on Build GRCh38
rs762389791 Chr.7: 41965659-41965659 on Build GRCh38
rs776349164 Chr.7: 41965705-41965705 on Build GRCh38
rs761887854 Chr.7: 41965559-41965559 on Build GRCh38
rs769447723 Chr.7: 41965719-41965719 on Build GRCh38
rs748236819 Chr.7: 41965625-41965625 on Build GRCh38
rs756384373 Chr.7: 41965600-41965600 on Build GRCh38
rs746960838 Chr.7: 41965702-41965702 on Build GRCh38
rs761143091 Chr.7: 41965727-41965727 on Build GRCh38
rs774628471 Chr.7: 41965624-41965624 on Build GRCh38
rs756608797 Chr.7: 41965683-41965683 on Build GRCh38
rs377548569 Chr.7: 41965572-41965572 on Build GRCh38
rs996732022 Chr.7: 41965745-41965745 on Build GRCh38
rs773211288 Chr.7: 41965563-41965563 on Build GRCh38
rs780349828 Chr.7: 41965607-41965607 on Build GRCh38
rs760545738 Chr.7: 41965635-41965635 on Build GRCh38
rs116840768 Chr.7: 41965634-41965634 on Build GRCh38
rs143942705 Chr.7: 41965564-41965564 on Build GRCh38
rs755313334 Chr.7: 41965734-41965734 on Build GRCh38
rs116840769 Chr.7: 41965617-41965617 on Build GRCh38
rs116840770 Chr.7: 41965592-41965592 on Build GRCh38
rs758667297 Chr.7: 41965748-41965748 on Build GRCh38
rs375822737 Chr.7: 41965641-41965641 on Build GRCh38
rs764694374 Chr.7: 41965566-41965566 on Build GRCh38
rs752206282 Chr.7: 41965658-41965658 on Build GRCh38
rs780328643 Chr.7: 41965692-41965692 on Build GRCh38
rs368061722 Chr.7: 41965722-41965722 on Build GRCh38
rs116840767 Chr.7: 41965687-41965687 on Build GRCh38
rs768034418 Chr.7: 41965676-41965676 on Build GRCh38
rs770879979 Chr.7: 41965704-41965704 on Build GRCh38
rs552755211 Chr.7: 41965730-41965730 on Build GRCh38
rs530155155 Chr.7: 41965644-41965644 on Build GRCh38
rs1048260302 Chr.7: 41965754-41965754 on Build GRCh38
rs767546154 Chr.7: 41965560-41965560 on Build GRCh38
rs772233504 Chr.7: 41965700-41965700 on Build GRCh38
rs781610968 Chr.7: 41965584-41965584 on Build GRCh38
rs776456548 Chr.7: 41965638-41965638 on Build GRCh38
rs896103474 Chr.7: 41965662-41965662 on Build GRCh38
rs374088219 Chr.7: 41965718-41965718 on Build GRCh38
rs542238121 Chr.7: 41965701-41965701 on Build GRCh38
rs778051402 Chr.7: 41965753-41965753 on Build GRCh38
rs370676430 Chr.7: 41965694-41965694 on Build GRCh38
rs750890276 Chr.7: 41965681-41965681 on Build GRCh38
rs116840766 Chr.7: 41965749-41965749 on Build GRCh38
rs878976775 Chr.7: 41965619-41965619 on Build GRCh38
rs1053587562 Chr.7: 41965677-41965677 on Build GRCh38
rs753384480 Chr.7: 41965565-41965565 on Build GRCh38
rs373688469 Chr.7: 41965602-41965602 on Build GRCh38
rs779300306 Chr.7: 41965738-41965738 on Build GRCh38
rs867889032 Chr.7: 41965688-41965688 on Build GRCh38
rs948927518 Chr.7: 41965606-41965606 on Build GRCh38
rs281864935 Chr.7: 41965686-41965687 on Build GRCh38
rs746243124 Chr.7: 41965596-41965596 on Build GRCh38
rs752928283 Chr.7: 41965744-41965744 on Build GRCh38
rs772143453 Chr.7: 41965632-41965632 on Build GRCh38
rs779212391 Chr.7: 41965695-41965695 on Build GRCh38

View Full Product Details