Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFAP2E
Entrez Gene ID
339488
Gene Name
transcription factor AP-2 epsilon
Gene Aliases
AP2E
Chromosome Location
Chr.1: 35571067 - 35595591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178548.3 NP_848643.2 5 - 6 1114 80
XM_017001139.1 XP_016856628.1 5 - 6 3417 80
GenBank mRNA AY326454.1 - 5 - 6 1114 80
BC041175.1 - 5 - 6 1114 80

View Full Product Details