Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79835125-79835350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX4
Entrez Gene ID
8535
Gene Name
chromobox 4
Gene Aliases
NBP16, PC2
UniGene
Hs.405046
Gene Chromosome Location
Chr.17: 79833156-79839414 on Build GRCh38
Transcripts
NM_003655.2 XM_011525399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4621429 c.457C>G p.Q153E Chr.17: 79835185-79835185 on Build GRCh38
6046405 c.432G>A p.P144P Chr.17: 79835210-79835210 on Build GRCh38
6621035 c.409C>T p.R137W Chr.17: 79835233-79835233 on Build GRCh38
6148485 c.447G>C p.K149N Chr.17: 79835195-79835195 on Build GRCh38
3672610 c.342G>A p.K114K Chr.17: 79835300-79835300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748919497 Chr.17: 79835199-79835199 on Build GRCh38
rs541220750 Chr.17: 79835246-79835246 on Build GRCh38
rs373367816 Chr.17: 79835229-79835229 on Build GRCh38
rs865831023 Chr.17: 79835279-79835279 on Build GRCh38
rs370792646 Chr.17: 79835221-79835221 on Build GRCh38
rs542359767 Chr.17: 79835292-79835292 on Build GRCh38
rs190305993 Chr.17: 79835243-79835243 on Build GRCh38
rs747829221 Chr.17: 79835178-79835178 on Build GRCh38
rs377573323 Chr.17: 79835331-79835331 on Build GRCh38
rs966035963 Chr.17: 79835281-79835281 on Build GRCh38
rs1048843257 Chr.17: 79835218-79835218 on Build GRCh38
rs1004564609 Chr.17: 79835258-79835258 on Build GRCh38
rs887539557 Chr.17: 79835224-79835224 on Build GRCh38
rs11650229 Chr.17: 79835259-79835259 on Build GRCh38
rs61737009 Chr.17: 79835162-79835162 on Build GRCh38
rs751687189 Chr.17: 79835160-79835160 on Build GRCh38
rs759665188 Chr.17: 79835312-79835312 on Build GRCh38
rs765846377 Chr.17: 79835209-79835210 on Build GRCh38
rs201600717 Chr.17: 79835186-79835186 on Build GRCh38
rs142055924 Chr.17: 79835309-79835309 on Build GRCh38
rs770739316 Chr.17: 79835217-79835217 on Build GRCh38
rs745400849 Chr.17: 79835222-79835222 on Build GRCh38
rs777761894 Chr.17: 79835198-79835198 on Build GRCh38
rs767557503 Chr.17: 79835315-79835315 on Build GRCh38
rs766410818 Chr.17: 79835150-79835150 on Build GRCh38
rs777194073 Chr.17: 79835232-79835232 on Build GRCh38

View Full Product Details