The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286259.1 | NP_001273188.1 | 2 - 3 | 1346 | 123 |
NM_182503.2 | NP_872309.2 | 2 - 3 | 248 | 123 | |
XM_011535439.2 | XP_011533741.1 | 5 - 6 | 404 | 123 | |
XM_011535440.1 | XP_011533742.1 | 5 - 6 | 3136 | 123 | |
XM_011535444.2 | XP_011533746.1 | 4 - 5 | 267 | 123 | |
XM_017010260.1 | XP_016865749.1 | 3 - 4 | 3665 | 123 | |
XM_017010261.1 | XP_016865750.1 | 2 - 3 | 3622 | 123 | |
XM_017010262.1 | XP_016865751.1 | 2 - 3 | 126 | 123 | |
GenBank mRNA | AK126201.1 | - | 2 - 3 | 1352 | 123 |
BC037955.1 | - | 2 - 3 | 208 | 123 | |
BX538182.1 | - | 2 - 3 | 646 | 123 |