Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDDC1
Entrez Gene ID
347862
Gene Name
Parkinson disease 7 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 767095 - 777502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318818.1 NP_001305747.1 6 - 7 624 120
NM_001318824.1 NP_001305753.1 6 - 7 593 120
NM_182612.3 NP_872418.1 7 - 8 701 120
NR_134867.1 6 - 7 732 120
NR_134868.1 7 - 8 795 120
GenBank mRNA AK091602.1 - 7 - 8 683 120
AK092816.1 - 6 - 7 578 120
AK293559.1 - 6 - 7 609 120
BC112145.1 - 7 - 8 682 120
BC143419.1 - 6 - 7 605 120
BC143420.1 - 6 - 7 574 120
BC143421.1 - 7 - 8 776 120
BC143423.1 - 6 - 7 713 120

View Full Product Details