The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003806.2 | NP_003797.1 | 1 - 1 | 354 | 156 |
NR_073189.1 | 1 - 1 | 354 | 156 | ||
XM_017020146.1 | XP_016875635.1 | 1 - 1 | 350 | 156 | |
GenBank mRNA | U76376.1 | - | 1 - 1 | 354 | 156 |