The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272071.1 | NP_001259000.1 | 6 - 6 | 1677 | 142 |
NM_003916.4 | NP_003907.3 | 5 - 5 | 1668 | 142 | |
GenBank mRNA | AF251295.1 | - | 5 - 5 | 1422 | 142 |
AK125525.1 | - | 1 - 1 | 2819 | 142 | |
AK130075.1 | - | 5 - 5 | 1470 | 142 | |
BX537780.1 | - | 5 - 5 | 1362 | 142 |