The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167820.1 | NP_001161292.1 | 3 - 3 | 747 | 85 |
NM_001167821.1 | NP_001161293.1 | 2 - 2 | 577 | 85 | |
NM_001167822.2 | NP_001161294.1 | 2 - 2 | 634 | 85 | |
NM_001167823.1 | NP_001161295.1 | 2 - 2 | 650 | 85 | |
NM_001317074.1 | NP_001304003.1 | 2 - 2 | 627 | 85 | |
NM_001317075.1 | NP_001304004.1 | 2 - 2 | 543 | 85 | |
NM_006698.3 | NP_006689.1 | 2 - 2 | 614 | 85 | |
XM_017027599.1 | XP_016883088.1 | 2 - 2 | 1527 | 85 | |
GenBank mRNA | AF053470.1 | - | 2 - 2 | 553 | 85 |
AK124822.1 | - | 3 - 3 | 472 | 85 | |
AL049288.1 | - | 2 - 2 | 560 | 85 | |
BC047692.1 | - | 2 - 2 | 545 | 85 | |
BC071704.1 | - | 2 - 2 | 539 | 85 |