Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41746141-41746368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4714162 c.544G>A p.D182N Chr.4: 41746208-41746208 on Build GRCh38
3603895 c.561C>T p.A187A Chr.4: 41746191-41746191 on Build GRCh38
3603894 c.609C>T p.N203N Chr.4: 41746143-41746143 on Build GRCh38
1745539 c.490_491insG p.A164fs*14 Chr.4: 41746261-41746262 on Build GRCh38
4625305 c.497C>T p.A166V Chr.4: 41746255-41746255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752437432 Chr.4: 41746272-41746272 on Build GRCh38
rs104893856 Chr.4: 41746162-41746162 on Build GRCh38
rs758358906 Chr.4: 41746287-41746287 on Build GRCh38
rs746303997 Chr.4: 41746221-41746221 on Build GRCh38
rs751132977 Chr.4: 41746327-41746327 on Build GRCh38
rs534384164 Chr.4: 41746271-41746271 on Build GRCh38
rs980457183 Chr.4: 41746328-41746328 on Build GRCh38
rs865798137 Chr.4: 41746212-41746212 on Build GRCh38
rs780967053 Chr.4: 41746342-41746342 on Build GRCh38
rs547677836 Chr.4: 41746266-41746266 on Build GRCh38
rs894529129 Chr.4: 41746292-41746292 on Build GRCh38
rs770661130 Chr.4: 41746203-41746203 on Build GRCh38
rs17885216 Chr.4: 41746200-41746200 on Build GRCh38
rs756712862 Chr.4: 41746335-41746335 on Build GRCh38
rs774521395 Chr.4: 41746255-41746255 on Build GRCh38
rs770281540 Chr.4: 41746238-41746238 on Build GRCh38
rs931109316 Chr.4: 41746248-41746248 on Build GRCh38
rs745523448 Chr.4: 41746347-41746347 on Build GRCh38
rs767837376 Chr.4: 41746265-41746265 on Build GRCh38
rs775931264 Chr.4: 41746244-41746244 on Build GRCh38
rs763569681 Chr.4: 41746249-41746249 on Build GRCh38
rs780690330 Chr.4: 41746209-41746209 on Build GRCh38
rs761776635 Chr.4: 41746260-41746260 on Build GRCh38
rs866814213 Chr.4: 41746234-41746234 on Build GRCh38
rs144414806 Chr.4: 41746161-41746161 on Build GRCh38
rs769236699 Chr.4: 41746254-41746254 on Build GRCh38
rs17881486 Chr.4: 41746302-41746302 on Build GRCh38
rs765027210 Chr.4: 41746269-41746269 on Build GRCh38

View Full Product Details