Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHLDA1
Entrez Gene ID
22822
Gene Name
pleckstrin homology like domain family A member 1
Gene Aliases
DT1P1B11, PHRIP, TDAG51
Chromosome Location
Chr.12: 76025447 - 76031776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007350.3 NP_031376.3 1 - 1 1129 78
GenBank mRNA AF220656.1 - 1 - 1 48 78
AK304224.1 - 2 - 2 941 78
BC018929.2 - 1 - 1 937 78
BC110820.1 - 1 - 1 928 78
BC126425.1 - 1 - 1 828 78
BC130427.1 - 1 - 1 828 78
Z50194.1 - 1 - 1 1250 78

View Full Product Details