The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300783.1 | NP_001287712.1 | 2 - 2 | 1074 | 83 |
NM_001308087.1 | NP_001295016.1 | 2 - 2 | 787 | 83 | |
NM_001308088.1 | NP_001295017.1 | 4 - 4 | 1027 | 83 | |
NM_016644.2 | NP_057728.1 | 3 - 3 | 1199 | 83 | |
XM_011543452.2 | XP_011541754.1 | 3 - 3 | 2686 | 83 | |
XM_011543453.2 | XP_011541755.1 | 3 - 3 | 1168 | 83 | |
XM_011543454.2 | XP_011541756.1 | 4 - 4 | 1274 | 83 | |
GenBank mRNA | AF242769.1 | - | 3 - 3 | 1153 | 83 |
BC038838.1 | - | 3 - 3 | 886 | 83 | |
BC092474.1 | - | 2 - 2 | 1075 | 83 |