The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178434.2 | NP_848521.1 | 1 - 1 | 220 | 69 |
GenBank mRNA | BC130365.1 | - | 1 - 1 | 220 | 69 |
BC130367.1 | - | 1 - 1 | 220 | 69 |