Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRAM2
Entrez Gene ID
128338
Gene Name
DNA damage regulated autophagy modulator 2
Gene Aliases
CORD21, PRO180, TMEM77, WWFQ154
Chromosome Location
Chr.1: 111117332 - 111140216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178454.4 NP_848549.3 9 - 9 1105 140
XM_005270469.2 XP_005270526.1 10 - 10 1163 140
XM_005270470.1 XP_005270527.1 10 - 10 1286 140
XM_006710361.1 XP_006710424.1 10 - 10 1123 140
XM_006710362.1 XP_006710425.1 10 - 10 1096 140
XM_011540707.1 XP_011539009.1 10 - 10 1104 140
XM_011540708.1 XP_011539010.1 10 - 10 1184 140
XM_017000320.1 XP_016855809.1 9 - 9 1047 140
XM_017000321.1 XP_016855810.1 9 - 9 906 140
GenBank mRNA AK075350.1 - 5 - 5 551 140
AL833098.1 - 9 - 9 1473 140
AY336747.1 - 9 - 9 983 140
AY358492.1 - 9 - 9 968 140
BC047025.1 - 8 - 8 920 140
BC091509.1 - 10 - 10 1197 140

View Full Product Details