The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178460.2 | NP_848555.2 | 2 - 2 | 352 | 59 |
GenBank mRNA | AK093083.1 | - | 2 - 2 | 1498 | 59 |
BC033502.1 | - | 2 - 2 | 352 | 59 |