Product Details

Assay Reference Genome
Location

Chr.19:40403762 on build GRCh38
Cytoband
19q13.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
57716
Gene Symbol
PRX
Gene Name
periaxin
Gene Aliases
CMT4F
Location
Chr.19:40393762-40414718 on build GRCh38
Assay Gene Location
Within Exon 6
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PRX NM_020956.2 5 397 NP_066007.1
NM_181882.2 5 397 NP_870998.2
XM_011527171.2 5 389 XP_011525473.1
AB046840.1 1 527 BAB13446.1
AF321191.1 5 397 AAK19279.1
AF321192.1 5 397 AAK19280.1
BC067266.1 5 399 AAH67266.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833831 Chr19:40290833 - 40454958 on Build GRCh38 Loss BLVRB SERTAD3 MIR6796 PLD3 C19orf47 PRX LOC105372489 HIPK4 SERTAD1

View Full Product Details