Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 118236743-118236970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD7
Entrez Gene ID
56311
Gene Name
ankyrin repeat domain 7
Gene Aliases
TSA806
UniGene
Hs.729340
Gene Chromosome Location
Chr.7: 118224658-118242731 on Build GRCh38
Transcripts
NM_019644.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3257489 c.657C>A p.V219V Chr.7: 118236811-118236811 on Build GRCh38
5798533 c.772+1G>A p.? Chr.7: 118236927-118236927 on Build GRCh38
3257493 c.734C>A p.S245* Chr.7: 118236888-118236888 on Build GRCh38
744899 c.763G>A p.V255I Chr.7: 118236917-118236917 on Build GRCh38
3257491 c.672A>T p.P224P Chr.7: 118236826-118236826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774762891 Chr.7: 118236813-118236813 on Build GRCh38
rs766691129 Chr.7: 118236883-118236883 on Build GRCh38
rs775755871 Chr.7: 118236775-118236776 on Build GRCh38
rs1039398407 Chr.7: 118236810-118236810 on Build GRCh38
rs774523105 Chr.7: 118236904-118236905 on Build GRCh38
rs892773192 Chr.7: 118236835-118236835 on Build GRCh38
rs769059448 Chr.7: 118236808-118236808 on Build GRCh38
rs377637661 Chr.7: 118236903-118236903 on Build GRCh38
rs775845687 Chr.7: 118236797-118236797 on Build GRCh38
rs200463335 Chr.7: 118236872-118236872 on Build GRCh38
rs745889886 Chr.7: 118236926-118236926 on Build GRCh38
rs751470762 Chr.7: 118236882-118236882 on Build GRCh38
rs902555813 Chr.7: 118236871-118236871 on Build GRCh38
rs762360582 Chr.7: 118236821-118236821 on Build GRCh38
rs766011037 Chr.7: 118236836-118236836 on Build GRCh38
rs763074069 Chr.7: 118236783-118236783 on Build GRCh38
rs370006498 Chr.7: 118236913-118236913 on Build GRCh38
rs889887004 Chr.7: 118236777-118236778 on Build GRCh38
rs768665754 Chr.7: 118236843-118236843 on Build GRCh38
rs1043728328 Chr.7: 118236792-118236792 on Build GRCh38
rs752385461 Chr.7: 118236871-118236871 on Build GRCh38
rs759270540 Chr.7: 118236845-118236845 on Build GRCh38
rs764928734 Chr.7: 118236869-118236869 on Build GRCh38
rs763924668 Chr.7: 118236881-118236881 on Build GRCh38
rs570626299 Chr.7: 118236811-118236811 on Build GRCh38
rs781249866 Chr.7: 118236915-118236915 on Build GRCh38
rs749672166 Chr.7: 118236803-118236803 on Build GRCh38
rs776242086 Chr.7: 118236841-118236841 on Build GRCh38
rs113033396 Chr.7: 118236927-118236927 on Build GRCh38

View Full Product Details