Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109626587-109626820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD2
Entrez Gene ID
271
Gene Name
adenosine monophosphate deaminase 2
Gene Aliases
PCH9, SPG63
UniGene
Hs.82927
Gene Chromosome Location
Chr.1: 109619813-109632055 on Build GRCh38
Transcripts
NM_001257360.1 NM_001257361.1 NM_001308170.1 NM_004037.7 NM_139156.3 NM_203404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106329 c.698C>A p.P233Q Chr.1: 109626730-109626730 on Build GRCh38
7106328 c.341C>A p.P114Q Chr.1: 109626730-109626730 on Build GRCh38
6599399 c.734T>C p.V245A Chr.1: 109626766-109626766 on Build GRCh38
215446 c.751C>T p.R251W Chr.1: 109626783-109626783 on Build GRCh38
4970258 c.698C>T p.P233L Chr.1: 109626730-109626730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748214358 Chr.1: 109626710-109626710 on Build GRCh38
rs762910071 Chr.1: 109626768-109626768 on Build GRCh38
rs777990790 Chr.1: 109626800-109626800 on Build GRCh38
rs567213314 Chr.1: 109626618-109626618 on Build GRCh38
rs1018565703 Chr.1: 109626665-109626665 on Build GRCh38
rs1020294315 Chr.1: 109626635-109626635 on Build GRCh38
rs201603567 Chr.1: 109626749-109626749 on Build GRCh38
rs780772416 Chr.1: 109626780-109626780 on Build GRCh38
rs144177097 Chr.1: 109626776-109626776 on Build GRCh38
rs371797266 Chr.1: 109626707-109626707 on Build GRCh38
rs891094950 Chr.1: 109626637-109626637 on Build GRCh38
rs773453238 Chr.1: 109626718-109626718 on Build GRCh38
rs41314005 Chr.1: 109626708-109626708 on Build GRCh38
rs751043420 Chr.1: 109626774-109626774 on Build GRCh38
rs1008495194 Chr.1: 109626641-109626641 on Build GRCh38
rs752584321 Chr.1: 109626766-109626766 on Build GRCh38
rs552559905 Chr.1: 109626797-109626797 on Build GRCh38
rs779440090 Chr.1: 109626793-109626793 on Build GRCh38
rs190168259 Chr.1: 109626634-109626634 on Build GRCh38
rs755344801 Chr.1: 109626783-109626783 on Build GRCh38
rs772225396 Chr.1: 109626714-109626714 on Build GRCh38
rs748621330 Chr.1: 109626796-109626796 on Build GRCh38
rs567095077 Chr.1: 109626730-109626730 on Build GRCh38
rs991126362 Chr.1: 109626658-109626658 on Build GRCh38
rs776483432 Chr.1: 109626731-109626731 on Build GRCh38
rs749670390 Chr.1: 109626693-109626693 on Build GRCh38
rs550556212 Chr.1: 109626617-109626617 on Build GRCh38
rs772026550 Chr.1: 109626676-109626677 on Build GRCh38
rs183170041 Chr.1: 109626773-109626773 on Build GRCh38
rs765137918 Chr.1: 109626764-109626764 on Build GRCh38
rs960676448 Chr.1: 109626641-109626641 on Build GRCh38
rs760896648 Chr.1: 109626723-109626723 on Build GRCh38

View Full Product Details