Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 38426851-38427090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
UniGene
Hs.592182
Gene Chromosome Location
Chr.7: 38383694-38631567 on Build GRCh38
Transcripts
NM_001635.3 NM_139316.2 XM_006715689.3 XM_006715690.3 XM_011515271.2 XM_017011995.1 XM_017011996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3703222 c.1210A>G p.T404A Chr.7: 38426959-38426959 on Build GRCh38
4875211 c.1195T>A p.S399T Chr.7: 38426974-38426974 on Build GRCh38
3703221 c.1210A>G p.T404A Chr.7: 38426959-38426959 on Build GRCh38
1089477 c.1195T>A p.S399T Chr.7: 38426974-38426974 on Build GRCh38
4523240 c.1205G>A p.G402E Chr.7: 38426964-38426964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756223032 Chr.7: 38427014-38427016 on Build GRCh38
rs187620602 Chr.7: 38426886-38426886 on Build GRCh38
rs758356182 Chr.7: 38426925-38426925 on Build GRCh38
rs78935089 Chr.7: 38427059-38427059 on Build GRCh38
rs758307115 Chr.7: 38426908-38426908 on Build GRCh38
rs752512125 Chr.7: 38426919-38426919 on Build GRCh38
rs746779442 Chr.7: 38426944-38426944 on Build GRCh38
rs775058908 Chr.7: 38427025-38427025 on Build GRCh38
rs757010717 Chr.7: 38426951-38426951 on Build GRCh38
rs374487341 Chr.7: 38426905-38426905 on Build GRCh38
rs957562036 Chr.7: 38426999-38426999 on Build GRCh38
rs772608635 Chr.7: 38427030-38427030 on Build GRCh38
rs1021285161 Chr.7: 38427046-38427046 on Build GRCh38
rs1033116778 Chr.7: 38426998-38426998 on Build GRCh38
rs74721180 Chr.7: 38427060-38427060 on Build GRCh38
rs968926150 Chr.7: 38426934-38426934 on Build GRCh38
rs371065137 Chr.7: 38426976-38426976 on Build GRCh38
rs769361183 Chr.7: 38427006-38427006 on Build GRCh38
rs769918839 Chr.7: 38426890-38426890 on Build GRCh38
rs777491039 Chr.7: 38426927-38426927 on Build GRCh38
rs748661419 Chr.7: 38427028-38427028 on Build GRCh38
rs750688683 Chr.7: 38427010-38427011 on Build GRCh38
rs775699955 Chr.7: 38427058-38427058 on Build GRCh38
rs368751891 Chr.7: 38426972-38426972 on Build GRCh38

View Full Product Details