Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130123725-130123974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APLP2
Entrez Gene ID
334
Gene Name
amyloid beta precursor like protein 2
Gene Aliases
APLP-2, APPH, APPL2, CDEBP
UniGene
Hs.370247
Gene Chromosome Location
Chr.11: 130069821-130144811 on Build GRCh38
Transcripts
NM_001142276.1 NM_001142277.1 NM_001142278.1 NM_001243299.1 NM_001642.2 NR_024515.1 NR_024516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925547 c.1090+8C>T p.? Chr.11: 130123787-130123787 on Build GRCh38
3719899 c.1077G>A p.V359V Chr.11: 130123766-130123766 on Build GRCh38
3445304 c.1040G>T p.R347M Chr.11: 130123729-130123729 on Build GRCh38
1978080 c.1085C>T p.A362V Chr.11: 130123774-130123774 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767265271 Chr.11: 130123776-130123776 on Build GRCh38
rs966370547 Chr.11: 130123913-130123913 on Build GRCh38
rs527689416 Chr.11: 130123851-130123851 on Build GRCh38
rs752399273 Chr.11: 130123827-130123827 on Build GRCh38
rs758568357 Chr.11: 130123795-130123795 on Build GRCh38
rs546391563 Chr.11: 130123793-130123793 on Build GRCh38
rs751481974 Chr.11: 130123768-130123768 on Build GRCh38
rs1006301004 Chr.11: 130123888-130123888 on Build GRCh38
rs796527462 Chr.11: 130123850-130123850 on Build GRCh38
rs766245157 Chr.11: 130123763-130123763 on Build GRCh38
rs764964482 Chr.11: 130123757-130123757 on Build GRCh38
rs577335648 Chr.11: 130123758-130123758 on Build GRCh38
rs747515478 Chr.11: 130123800-130123800 on Build GRCh38
rs916924531 Chr.11: 130123761-130123761 on Build GRCh38
rs769264332 Chr.11: 130123814-130123814 on Build GRCh38
rs773887845 Chr.11: 130123821-130123821 on Build GRCh38
rs767468024 Chr.11: 130123823-130123823 on Build GRCh38
rs201772983 Chr.11: 130123804-130123804 on Build GRCh38
rs78611322 Chr.11: 130123928-130123928 on Build GRCh38
rs949724035 Chr.11: 130123772-130123772 on Build GRCh38
rs1010670260 Chr.11: 130123895-130123895 on Build GRCh38
rs375624657 Chr.11: 130123775-130123775 on Build GRCh38
rs369687086 Chr.11: 130123816-130123816 on Build GRCh38
rs752582093 Chr.11: 130123777-130123777 on Build GRCh38
rs372840531 Chr.11: 130123932-130123932 on Build GRCh38
rs759443645 Chr.11: 130123822-130123822 on Build GRCh38
rs113352584 Chr.11: 130123780-130123780 on Build GRCh38
rs372558282 Chr.11: 130123818-130123818 on Build GRCh38
rs115641832 Chr.11: 130123876-130123876 on Build GRCh38
rs959414642 Chr.11: 130123883-130123883 on Build GRCh38
rs762569937 Chr.11: 130123817-130123817 on Build GRCh38
rs369478296 Chr.11: 130123787-130123787 on Build GRCh38
rs1022031022 Chr.11: 130123902-130123902 on Build GRCh38
rs376905429 Chr.11: 130123830-130123830 on Build GRCh38
rs373556814 Chr.11: 130123794-130123794 on Build GRCh38
rs376424694 Chr.11: 130123808-130123808 on Build GRCh38
rs757376095 Chr.11: 130123785-130123785 on Build GRCh38
rs961217477 Chr.11: 130123898-130123898 on Build GRCh38
rs891956762 Chr.11: 130123809-130123809 on Build GRCh38
rs766585889 Chr.11: 130123805-130123805 on Build GRCh38
rs1016805200 Chr.11: 130123897-130123897 on Build GRCh38
rs561786130 Chr.11: 130123833-130123833 on Build GRCh38
rs1002151768 Chr.11: 130123868-130123868 on Build GRCh38

View Full Product Details