Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP2A7
- Entrez Gene ID
-
1549
- Gene Name
- cytochrome P450 family 2 subfamily A member 7
- Gene Aliases
-
CPA7, CPAD, CYP2A, CYPIIA7, P450-IIA4
- Chromosome Location
-
Chr.19: 40875439 - 40885952 on Build GRCh38
- Gene Symbol
- CYP2A7P1
- Entrez Gene ID
-
1550
- Gene Name
- cytochrome P450 family 2 subfamily A member 7 pseudogene 1
- Gene Aliases
-
CYP2A18PC, CYP2A18PN, CYP2A7P2, CYP2A7PT
- Chromosome Location
-
Chr.19: 41024251 - 41028263 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U22030.1 |
- |
1 - 1 |
370 |
67 |
U22044.1 |
- |
1 - 1 |
370 |
67 |
View Full Product Details