Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35042364-35042589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf131
Entrez Gene ID
138724
Gene Name
chromosome 9 open reading frame 131
Gene Aliases
-
UniGene
Hs.148250
Gene Chromosome Location
Chr.9: 35041095-35045991 on Build GRCh38
Transcripts
NM_001040410.2 NM_001040411.2 NM_001040412.2 NM_001287391.1 NM_203299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5566546 c.391C>A p.P131T Chr.9: 35042437-35042437 on Build GRCh38
1108545 c.143T>C p.F48S Chr.9: 35042397-35042397 on Build GRCh38
5566544 c.183G>T p.R61S Chr.9: 35042437-35042437 on Build GRCh38
1132500 c.187C>T p.R63* Chr.9: 35042441-35042441 on Build GRCh38
5566545 c.39G>T p.R13S Chr.9: 35042437-35042437 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535524374 Chr.9: 35042545-35042545 on Build GRCh38
rs35629439 Chr.9: 35042418-35042418 on Build GRCh38
rs757393091 Chr.9: 35042420-35042420 on Build GRCh38
rs758697383 Chr.9: 35042428-35042428 on Build GRCh38
rs990039598 Chr.9: 35042540-35042540 on Build GRCh38
rs756113619 Chr.9: 35042401-35042401 on Build GRCh38
rs753840149 Chr.9: 35042514-35042514 on Build GRCh38
rs780301099 Chr.9: 35042552-35042552 on Build GRCh38
rs773813587 Chr.9: 35042471-35042471 on Build GRCh38
rs747429941 Chr.9: 35042442-35042442 on Build GRCh38
rs767343512 Chr.9: 35042477-35042477 on Build GRCh38
rs777909205 Chr.9: 35042415-35042415 on Build GRCh38
rs770493312 Chr.9: 35042463-35042463 on Build GRCh38
rs760589456 Chr.9: 35042487-35042487 on Build GRCh38
rs376416549 Chr.9: 35042398-35042398 on Build GRCh38
rs144687989 Chr.9: 35042441-35042441 on Build GRCh38
rs777234897 Chr.9: 35042456-35042456 on Build GRCh38
rs935609197 Chr.9: 35042532-35042532 on Build GRCh38
rs149562005 Chr.9: 35042399-35042399 on Build GRCh38
rs370990455 Chr.9: 35042498-35042498 on Build GRCh38
rs546933723 Chr.9: 35042396-35042396 on Build GRCh38
rs752611739 Chr.9: 35042400-35042400 on Build GRCh38
rs555477612 Chr.9: 35042555-35042555 on Build GRCh38
rs775405723 Chr.9: 35042482-35042482 on Build GRCh38
rs759264116 Chr.9: 35042474-35042474 on Build GRCh38
rs140803500 Chr.9: 35042424-35042424 on Build GRCh38
rs373148082 Chr.9: 35042511-35042511 on Build GRCh38
rs763197415 Chr.9: 35042509-35042510 on Build GRCh38
rs746115889 Chr.9: 35042427-35042427 on Build GRCh38
rs377071885 Chr.9: 35042446-35042446 on Build GRCh38
rs370487591 Chr.9: 35042535-35042535 on Build GRCh38
rs764585077 Chr.9: 35042561-35042561 on Build GRCh38
rs570076649 Chr.9: 35042389-35042389 on Build GRCh38
rs765363833 Chr.9: 35042521-35042521 on Build GRCh38
rs569738572 Chr.9: 35042537-35042537 on Build GRCh38
rs748706565 Chr.9: 35042461-35042461 on Build GRCh38
rs978740434 Chr.9: 35042432-35042432 on Build GRCh38

View Full Product Details