Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137010597-137010830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA2
Entrez Gene ID
20
Gene Name
ATP binding cassette subfamily A member 2
Gene Aliases
ABC2
UniGene
Hs.421202
Gene Chromosome Location
Chr.9: 137007234-137028922 on Build GRCh38
Transcripts
NM_001606.4 NM_212533.2 XM_006716996.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3088326 c.6099C>T p.D2033D Chr.9: 137010695-137010695 on Build GRCh38
6589657 c.6119G>A p.R2040Q Chr.9: 137010675-137010675 on Build GRCh38
5663880 c.6128G>T p.R2043L Chr.9: 137010666-137010666 on Build GRCh38
3088327 c.6067G>T p.V2023L Chr.9: 137010727-137010727 on Build GRCh38
5003265 c.6097G>T p.D2033Y Chr.9: 137010697-137010697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777826954 Chr.9: 137010751-137010751 on Build GRCh38
rs748107138 Chr.9: 137010669-137010669 on Build GRCh38
rs761049038 Chr.9: 137010788-137010788 on Build GRCh38
rs202088446 Chr.9: 137010665-137010665 on Build GRCh38
rs760288576 Chr.9: 137010721-137010721 on Build GRCh38
rs201228361 Chr.9: 137010725-137010725 on Build GRCh38
rs112887513 Chr.9: 137010689-137010689 on Build GRCh38
rs762366113 Chr.9: 137010634-137010634 on Build GRCh38
rs369022993 Chr.9: 137010736-137010736 on Build GRCh38
rs920903771 Chr.9: 137010639-137010639 on Build GRCh38
rs749539909 Chr.9: 137010771-137010771 on Build GRCh38
rs774683678 Chr.9: 137010776-137010776 on Build GRCh38
rs964978809 Chr.9: 137010626-137010626 on Build GRCh38
rs752533142 Chr.9: 137010743-137010743 on Build GRCh38
rs377050255 Chr.9: 137010678-137010678 on Build GRCh38
rs779954816 Chr.9: 137010763-137010763 on Build GRCh38
rs374099561 Chr.9: 137010659-137010659 on Build GRCh38
rs769048305 Chr.9: 137010772-137010772 on Build GRCh38
rs762245502 Chr.9: 137010704-137010704 on Build GRCh38
rs753560193 Chr.9: 137010730-137010730 on Build GRCh38
rs975059896 Chr.9: 137010635-137010635 on Build GRCh38
rs772050092 Chr.9: 137010676-137010676 on Build GRCh38
rs768178321 Chr.9: 137010706-137010706 on Build GRCh38
rs754774965 Chr.9: 137010667-137010667 on Build GRCh38
rs775791084 Chr.9: 137010682-137010682 on Build GRCh38
rs778640238 Chr.9: 137010668-137010668 on Build GRCh38
rs763016030 Chr.9: 137010685-137010685 on Build GRCh38
rs187721371 Chr.9: 137010718-137010718 on Build GRCh38
rs754576760 Chr.9: 137010735-137010735 on Build GRCh38
rs200927243 Chr.9: 137010695-137010695 on Build GRCh38
rs773775153 Chr.9: 137010786-137010786 on Build GRCh38
rs758189176 Chr.9: 137010745-137010745 on Build GRCh38
rs370257727 Chr.9: 137010655-137010655 on Build GRCh38
rs530351497 Chr.9: 137010805-137010805 on Build GRCh38
rs748588348 Chr.9: 137010777-137010777 on Build GRCh38
rs769752127 Chr.9: 137010681-137010681 on Build GRCh38
rs372494989 Chr.9: 137010677-137010677 on Build GRCh38
rs772309931 Chr.9: 137010780-137010780 on Build GRCh38
rs371774563 Chr.9: 137010753-137010753 on Build GRCh38
rs749999145 Chr.9: 137010656-137010656 on Build GRCh38
rs753376326 Chr.9: 137010666-137010666 on Build GRCh38
rs375404413 Chr.9: 137010761-137010761 on Build GRCh38

View Full Product Details