Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 427945-428206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
UniGene
Hs.501622
Gene Chromosome Location
Chr.11: 417930-442011 on Build GRCh38
Transcripts
NM_001012302.2 XM_011520048.1 XM_011520051.2 XM_011520052.2 XM_011520053.2 XM_017017644.1 XM_017017645.1 XM_017017646.1 XM_017017647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6189537 c.1334G>A p.R445K Chr.11: 428088-428088 on Build GRCh38
1746226 c.1241C>T p.S414L Chr.11: 428181-428181 on Build GRCh38
5422516 c.1334+6G>C p.? Chr.11: 428082-428082 on Build GRCh38
6518476 c.1275C>T p.F425F Chr.11: 428147-428147 on Build GRCh38
5004422 c.1247G>A p.R416Q Chr.11: 428175-428175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199877282 Chr.11: 428031-428031 on Build GRCh38
rs370870803 Chr.11: 428095-428095 on Build GRCh38
rs141612749 Chr.11: 428120-428120 on Build GRCh38
rs1022208890 Chr.11: 427983-427983 on Build GRCh38
rs763741740 Chr.11: 428049-428049 on Build GRCh38
rs779719871 Chr.11: 428060-428060 on Build GRCh38
rs747254082 Chr.11: 428169-428169 on Build GRCh38
rs754842224 Chr.11: 428144-428144 on Build GRCh38
rs746404674 Chr.11: 428081-428081 on Build GRCh38
rs368370867 Chr.11: 428080-428080 on Build GRCh38
rs182005096 Chr.11: 428025-428025 on Build GRCh38
rs776128506 Chr.11: 428187-428187 on Build GRCh38
rs775728151 Chr.11: 428092-428092 on Build GRCh38
rs560711468 Chr.11: 428077-428077 on Build GRCh38
rs545236900 Chr.11: 428013-428013 on Build GRCh38
rs573057747 Chr.11: 428012-428012 on Build GRCh38
rs1009020835 Chr.11: 428133-428133 on Build GRCh38
rs778126233 Chr.11: 428076-428076 on Build GRCh38
rs940940079 Chr.11: 428104-428104 on Build GRCh38
rs964966142 Chr.11: 428020-428020 on Build GRCh38
rs764357499 Chr.11: 428105-428105 on Build GRCh38
rs376901890 Chr.11: 428115-428115 on Build GRCh38
rs201806959 Chr.11: 428030-428030 on Build GRCh38
rs774184253 Chr.11: 428107-428107 on Build GRCh38
rs757430374 Chr.11: 428175-428175 on Build GRCh38
rs980520422 Chr.11: 427996-427996 on Build GRCh38
rs200822979 Chr.11: 428028-428028 on Build GRCh38
rs370309788 Chr.11: 428056-428056 on Build GRCh38
rs558654833 Chr.11: 428014-428014 on Build GRCh38
rs763430876 Chr.11: 428093-428093 on Build GRCh38
rs771048606 Chr.11: 428079-428079 on Build GRCh38
rs974974717 Chr.11: 428021-428021 on Build GRCh38
rs553257825 Chr.11: 428011-428011 on Build GRCh38
rs542936963 Chr.11: 428181-428181 on Build GRCh38
rs201974530 Chr.11: 428046-428046 on Build GRCh38
rs867140506 Chr.11: 427997-427997 on Build GRCh38
rs376095697 Chr.11: 428065-428065 on Build GRCh38
rs753717215 Chr.11: 428128-428128 on Build GRCh38
rs749182561 Chr.11: 428107-428108 on Build GRCh38
rs148485064 Chr.11: 428108-428108 on Build GRCh38
rs201483586 Chr.11: 428082-428082 on Build GRCh38
rs754718199 Chr.11: 428073-428073 on Build GRCh38
rs375996529 Chr.11: 428050-428050 on Build GRCh38
rs199668517 Chr.11: 428154-428154 on Build GRCh38
rs767171821 Chr.11: 428110-428110 on Build GRCh38
rs201008776 Chr.11: 428176-428176 on Build GRCh38
rs753064219 Chr.11: 428042-428042 on Build GRCh38
rs767100718 Chr.11: 428051-428051 on Build GRCh38
rs766399672 Chr.11: 428121-428121 on Build GRCh38

View Full Product Details