Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150228995-150229268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32E
Entrez Gene ID
81611
Gene Name
acidic nuclear phosphoprotein 32 family member E
Gene Aliases
LANP-L, LANPL
UniGene
Hs.656466
Gene Chromosome Location
Chr.1: 150218417-150236156 on Build GRCh38
Transcripts
NM_001136478.3 NM_001136479.2 NM_001280559.1 NM_001280560.1 NM_030920.4 XM_005245513.3 XM_005245514.3 XM_017002418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5461986 c.493+9T>G p.? Chr.1: 150229063-150229063 on Build GRCh38
4021937 c.377C>T p.T126I Chr.1: 150229188-150229188 on Build GRCh38
3704947 c.481G>T p.E161* Chr.1: 150229084-150229084 on Build GRCh38
6057930 c.469G>T p.D157Y Chr.1: 150229096-150229096 on Build GRCh38
4662105 c.467C>T p.P156L Chr.1: 150229098-150229098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369730881 Chr.1: 150229127-150229127 on Build GRCh38
rs782565601 Chr.1: 150229141-150229141 on Build GRCh38
rs782234067 Chr.1: 150229188-150229188 on Build GRCh38
rs782798771 Chr.1: 150229058-150229058 on Build GRCh38
rs782060801 Chr.1: 150229235-150229235 on Build GRCh38
rs782231779 Chr.1: 150229030-150229030 on Build GRCh38
rs782260989 Chr.1: 150229101-150229101 on Build GRCh38
rs782643352 Chr.1: 150229097-150229097 on Build GRCh38
rs782139113 Chr.1: 150229056-150229056 on Build GRCh38
rs782499585 Chr.1: 150229118-150229118 on Build GRCh38
rs782404279 Chr.1: 150229164-150229164 on Build GRCh38
rs782802339 Chr.1: 150229079-150229079 on Build GRCh38
rs587652193 Chr.1: 150229066-150229066 on Build GRCh38
rs781978711 Chr.1: 150229065-150229065 on Build GRCh38
rs782309466 Chr.1: 150229043-150229043 on Build GRCh38
rs587667046 Chr.1: 150229086-150229086 on Build GRCh38
rs151007873 Chr.1: 150229073-150229073 on Build GRCh38
rs782028923 Chr.1: 150229175-150229175 on Build GRCh38
rs932917381 Chr.1: 150229072-150229072 on Build GRCh38
rs144939904 Chr.1: 150229145-150229145 on Build GRCh38
rs940071447 Chr.1: 150229205-150229205 on Build GRCh38
rs781975027 Chr.1: 150229226-150229226 on Build GRCh38
rs782388606 Chr.1: 150229206-150229206 on Build GRCh38
rs781931705 Chr.1: 150229046-150229046 on Build GRCh38
rs781954026 Chr.1: 150229022-150229022 on Build GRCh38
rs587742318 Chr.1: 150229100-150229100 on Build GRCh38

View Full Product Details