Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73487721-73487948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFM
Entrez Gene ID
173
Gene Name
afamin
Gene Aliases
ALB2, ALBA, ALF
UniGene
Hs.168718
Gene Chromosome Location
Chr.4: 73481740-73504001 on Build GRCh38
Transcripts
NM_001133.2 XM_017007842.1 XM_017007843.1 XM_017007844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3605709 c.634T>A p.Y212N Chr.4: 73487742-73487742 on Build GRCh38
87974 c.643G>T p.A215S Chr.4: 73487751-73487751 on Build GRCh38
3605710 c.684G>A p.L228L Chr.4: 73487792-73487792 on Build GRCh38
1430930 c.674G>T p.G225V Chr.4: 73487782-73487782 on Build GRCh38
4660254 c.676G>A p.A226T Chr.4: 73487784-73487784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033636392 Chr.4: 73487861-73487861 on Build GRCh38
rs763407099 Chr.4: 73487765-73487765 on Build GRCh38
rs753026590 Chr.4: 73487758-73487758 on Build GRCh38
rs750166404 Chr.4: 73487787-73487787 on Build GRCh38
rs578007627 Chr.4: 73487923-73487923 on Build GRCh38
rs879202345 Chr.4: 73487890-73487890 on Build GRCh38
rs779479773 Chr.4: 73487805-73487805 on Build GRCh38
rs149715435 Chr.4: 73487799-73487799 on Build GRCh38
rs1053676781 Chr.4: 73487804-73487804 on Build GRCh38
rs771863777 Chr.4: 73487836-73487836 on Build GRCh38
rs1005408287 Chr.4: 73487908-73487908 on Build GRCh38
rs776832606 Chr.4: 73487842-73487842 on Build GRCh38
rs373816412 Chr.4: 73487849-73487849 on Build GRCh38
rs763114702 Chr.4: 73487873-73487873 on Build GRCh38
rs559545764 Chr.4: 73487887-73487887 on Build GRCh38
rs751922637 Chr.4: 73487779-73487779 on Build GRCh38
rs778893414 Chr.4: 73487845-73487845 on Build GRCh38
rs952725028 Chr.4: 73487882-73487882 on Build GRCh38
rs747883589 Chr.4: 73487825-73487825 on Build GRCh38
rs768775453 Chr.4: 73487805-73487805 on Build GRCh38
rs757707337 Chr.4: 73487780-73487780 on Build GRCh38
rs926580852 Chr.4: 73487772-73487772 on Build GRCh38
rs200258630 Chr.4: 73487822-73487822 on Build GRCh38
rs764689296 Chr.4: 73487771-73487771 on Build GRCh38
rs904059964 Chr.4: 73487867-73487867 on Build GRCh38
rs571635644 Chr.4: 73487919-73487919 on Build GRCh38
rs767212648 Chr.4: 73487782-73487782 on Build GRCh38
rs906169642 Chr.4: 73487846-73487846 on Build GRCh38
rs749220922 Chr.4: 73487820-73487820 on Build GRCh38

View Full Product Details