Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 15385393-15385620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGMO
Entrez Gene ID
392636
Gene Name
alkylglycerol monooxygenase
Gene Aliases
TMEM195
UniGene
Hs.670634
Gene Chromosome Location
Chr.7: 15117223-15562015 on Build GRCh38
Transcripts
NM_001004320.1 XM_006715730.1 XM_006715731.3 XM_011515402.2 XM_017012204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1087710 c.1031C>T p.A344V Chr.7: 15385489-15385489 on Build GRCh38
1698372 c.992C>T p.S331L Chr.7: 15385528-15385528 on Build GRCh38
6596288 c.986C>A p.S329* Chr.7: 15385534-15385534 on Build GRCh38
3832388 c.961A>G p.T321A Chr.7: 15385559-15385559 on Build GRCh38
402850 c.977C>A p.P326H Chr.7: 15385543-15385543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940526585 Chr.7: 15385445-15385445 on Build GRCh38
rs775191710 Chr.7: 15385565-15385565 on Build GRCh38
rs763654479 Chr.7: 15385522-15385522 on Build GRCh38
rs764361698 Chr.7: 15385505-15385506 on Build GRCh38
rs76269132 Chr.7: 15385435-15385435 on Build GRCh38
rs751479599 Chr.7: 15385525-15385525 on Build GRCh38
rs371899719 Chr.7: 15385590-15385590 on Build GRCh38
rs372369328 Chr.7: 15385498-15385498 on Build GRCh38
rs149623873 Chr.7: 15385562-15385562 on Build GRCh38
rs141075197 Chr.7: 15385503-15385503 on Build GRCh38
rs781025961 Chr.7: 15385535-15385535 on Build GRCh38
rs139340804 Chr.7: 15385509-15385509 on Build GRCh38
rs747012970 Chr.7: 15385466-15385466 on Build GRCh38
rs982863271 Chr.7: 15385539-15385539 on Build GRCh38
rs749443521 Chr.7: 15385478-15385478 on Build GRCh38
rs763566792 Chr.7: 15385572-15385572 on Build GRCh38
rs140207104 Chr.7: 15385543-15385543 on Build GRCh38
rs752197897 Chr.7: 15385450-15385450 on Build GRCh38
rs146651080 Chr.7: 15385511-15385511 on Build GRCh38
rs772284524 Chr.7: 15385494-15385494 on Build GRCh38
rs758155268 Chr.7: 15385503-15385504 on Build GRCh38
rs200122012 Chr.7: 15385475-15385475 on Build GRCh38
rs777617525 Chr.7: 15385453-15385453 on Build GRCh38
rs201562437 Chr.7: 15385556-15385556 on Build GRCh38
rs766906405 Chr.7: 15385439-15385439 on Build GRCh38
rs368430933 Chr.7: 15385575-15385575 on Build GRCh38
rs1055438899 Chr.7: 15385459-15385459 on Build GRCh38
rs199707904 Chr.7: 15385436-15385436 on Build GRCh38
rs757855092 Chr.7: 15385452-15385452 on Build GRCh38
rs754699109 Chr.7: 15385446-15385446 on Build GRCh38
rs770943190 Chr.7: 15385523-15385523 on Build GRCh38
rs771723699 Chr.7: 15385447-15385447 on Build GRCh38
rs150334002 Chr.7: 15385555-15385555 on Build GRCh38
rs561697174 Chr.7: 15385442-15385442 on Build GRCh38
rs1001638055 Chr.7: 15385457-15385457 on Build GRCh38
rs752467515 Chr.7: 15385455-15385455 on Build GRCh38
rs762646129 Chr.7: 15385570-15385570 on Build GRCh38
rs146527477 Chr.7: 15385487-15385487 on Build GRCh38
rs771391281 Chr.7: 15385508-15385508 on Build GRCh38
rs759873551 Chr.7: 15385560-15385560 on Build GRCh38
rs752295082 Chr.7: 15385512-15385512 on Build GRCh38
rs1010925531 Chr.7: 15385435-15385435 on Build GRCh38
rs137980424 Chr.7: 15385557-15385557 on Build GRCh38
rs892911481 Chr.7: 15385461-15385461 on Build GRCh38
rs576872902 Chr.7: 15385488-15385488 on Build GRCh38
rs533162418 Chr.7: 15385541-15385541 on Build GRCh38
rs549085660 Chr.7: 15385563-15385563 on Build GRCh38
rs773081953 Chr.7: 15385496-15385496 on Build GRCh38
rs562667253 Chr.7: 15385518-15385518 on Build GRCh38
rs972932289 Chr.7: 15385548-15385548 on Build GRCh38
rs144084616 Chr.7: 15385471-15385471 on Build GRCh38
rs768884011 Chr.7: 15385482-15385482 on Build GRCh38
rs772255699 Chr.7: 15385551-15385551 on Build GRCh38
rs890013165 Chr.7: 15385432-15385432 on Build GRCh38
rs764040888 Chr.7: 15385531-15385533 on Build GRCh38
rs375360314 Chr.7: 15385499-15385499 on Build GRCh38

View Full Product Details