Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46920362-46920623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP35
Entrez Gene ID
2909
Gene Name
Rho GTPase activating protein 35
Gene Aliases
GRF-1, GRLF1, P190-A, P190A, p190ARhoGAP, p190RhoGAP
UniGene
Hs.509447
Gene Chromosome Location
Chr.19: 46861002-47005077 on Build GRCh38
Transcripts
NM_004491.4 XM_011526873.2 XM_017026714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5464641 c.1850G>A p.R617Q Chr.19: 46920525-46920525 on Build GRCh38
5787370 c.1828C>T p.R610* Chr.19: 46920503-46920503 on Build GRCh38
7156406 c.1874A>G p.K625R Chr.19: 46920549-46920549 on Build GRCh38
5496661 c.1930G>A p.V644I Chr.19: 46920605-46920605 on Build GRCh38
7138123 c.1790G>C p.R597T Chr.19: 46920465-46920465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374629791 Chr.19: 46920538-46920538 on Build GRCh38
rs369018120 Chr.19: 46920400-46920400 on Build GRCh38
rs368789251 Chr.19: 46920426-46920426 on Build GRCh38
rs373252908 Chr.19: 46920404-46920404 on Build GRCh38
rs761965356 Chr.19: 46920417-46920417 on Build GRCh38
rs367600555 Chr.19: 46920571-46920571 on Build GRCh38
rs769392977 Chr.19: 46920393-46920393 on Build GRCh38
rs7250030 Chr.19: 46920418-46920418 on Build GRCh38
rs370275563 Chr.19: 46920577-46920577 on Build GRCh38
rs774301596 Chr.19: 46920405-46920405 on Build GRCh38
rs371130864 Chr.19: 46920508-46920508 on Build GRCh38
rs377726894 Chr.19: 46920493-46920493 on Build GRCh38
rs894375491 Chr.19: 46920444-46920444 on Build GRCh38
rs777872121 Chr.19: 46920591-46920591 on Build GRCh38
rs1063735 Chr.19: 46920524-46920524 on Build GRCh38
rs750559194 Chr.19: 46920425-46920425 on Build GRCh38
rs751915398 Chr.19: 46920443-46920443 on Build GRCh38
rs376565594 Chr.19: 46920414-46920414 on Build GRCh38
rs759394950 Chr.19: 46920550-46920550 on Build GRCh38
rs774215342 Chr.19: 46920547-46920547 on Build GRCh38
rs769146124 Chr.19: 46920501-46920501 on Build GRCh38
rs1017745324 Chr.19: 46920511-46920511 on Build GRCh38
rs781711227 Chr.19: 46920391-46920391 on Build GRCh38
rs551334120 Chr.19: 46920413-46920413 on Build GRCh38
rs375707432 Chr.19: 46920392-46920392 on Build GRCh38
rs755391551 Chr.19: 46920479-46920479 on Build GRCh38
rs769153230 Chr.19: 46920385-46920385 on Build GRCh38
rs200520064 Chr.19: 46920482-46920482 on Build GRCh38
rs762697644 Chr.19: 46920401-46920401 on Build GRCh38
rs748835273 Chr.19: 46920536-46920536 on Build GRCh38

View Full Product Details