Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114343621-114343894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
-
UniGene
Hs.494895
Gene Chromosome Location
Chr.9: 114334149-114398498 on Build GRCh38
Transcripts
NM_001317950.1 NM_001317952.1 NM_030767.5 XM_005252244.2 XM_005252245.1 XM_005252247.4 XM_005252248.2 XM_006717294.1 XM_011519063.2 XM_011519065.2 XM_017015172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3788152 c.3751G>T p.D1251Y Chr.9: 114343714-114343714 on Build GRCh38
6597907 c.3736C>T p.P1246S Chr.9: 114343729-114343729 on Build GRCh38
4878987 c.3666C>T p.H1222H Chr.9: 114343799-114343799 on Build GRCh38
5875404 c.3662-1G>A p.? Chr.9: 114343804-114343804 on Build GRCh38
4932706 c.3706A>G p.N1236D Chr.9: 114343759-114343759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373022948 Chr.9: 114343798-114343798 on Build GRCh38
rs763084560 Chr.9: 114343819-114343820 on Build GRCh38
rs751765388 Chr.9: 114343768-114343768 on Build GRCh38
rs376221512 Chr.9: 114343816-114343816 on Build GRCh38
rs772877978 Chr.9: 114343704-114343704 on Build GRCh38
rs142443528 Chr.9: 114343723-114343723 on Build GRCh38
rs1001315294 Chr.9: 114343766-114343766 on Build GRCh38
rs773932227 Chr.9: 114343713-114343713 on Build GRCh38
rs968655172 Chr.9: 114343736-114343736 on Build GRCh38
rs745557148 Chr.9: 114343808-114343808 on Build GRCh38
rs754792132 Chr.9: 114343657-114343657 on Build GRCh38
rs769803934 Chr.9: 114343689-114343689 on Build GRCh38
rs762965662 Chr.9: 114343758-114343758 on Build GRCh38
rs759542152 Chr.9: 114343772-114343772 on Build GRCh38
rs765696116 Chr.9: 114343740-114343740 on Build GRCh38
rs148801911 Chr.9: 114343709-114343709 on Build GRCh38
rs12000918 Chr.9: 114343861-114343861 on Build GRCh38
rs200169111 Chr.9: 114343732-114343732 on Build GRCh38
rs764995250 Chr.9: 114343822-114343822 on Build GRCh38
rs370887054 Chr.9: 114343688-114343688 on Build GRCh38
rs759222112 Chr.9: 114343731-114343731 on Build GRCh38
rs770720717 Chr.9: 114343710-114343710 on Build GRCh38
rs571710278 Chr.9: 114343850-114343850 on Build GRCh38
rs762251074 Chr.9: 114343739-114343739 on Build GRCh38
rs763415523 Chr.9: 114343844-114343844 on Build GRCh38
rs773771386 Chr.9: 114343840-114343840 on Build GRCh38
rs749224473 Chr.9: 114343780-114343780 on Build GRCh38
rs750509009 Chr.9: 114343752-114343752 on Build GRCh38
rs771707702 Chr.9: 114343809-114343809 on Build GRCh38
rs1052136548 Chr.9: 114343654-114343654 on Build GRCh38
rs983713558 Chr.9: 114343860-114343860 on Build GRCh38
rs755201054 Chr.9: 114343771-114343771 on Build GRCh38
rs777032921 Chr.9: 114343734-114343734 on Build GRCh38
rs766437297 Chr.9: 114343761-114343761 on Build GRCh38
rs556752187 Chr.9: 114343646-114343646 on Build GRCh38
rs1040270316 Chr.9: 114343746-114343746 on Build GRCh38
rs781005872 Chr.9: 114343664-114343664 on Build GRCh38
rs151319826 Chr.9: 114343773-114343773 on Build GRCh38
rs778455451 Chr.9: 114343799-114343799 on Build GRCh38
rs16928348 Chr.9: 114343856-114343856 on Build GRCh38
rs776894559 Chr.9: 114343815-114343815 on Build GRCh38
rs561306665 Chr.9: 114343777-114343777 on Build GRCh38

View Full Product Details