Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8109953-8110214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOXE3
Entrez Gene ID
59344
Gene Name
arachidonate lipoxygenase 3
Gene Aliases
ARCI3, E-LOX, eLOX-3, eLOX3
UniGene
Hs.232770
Gene Chromosome Location
Chr.17: 8095900-8118916 on Build GRCh38
Transcripts
NM_001165960.1 NM_021628.2 XM_017024921.1 XM_017024922.1 XM_017024923.1 XM_017024924.1 XM_017024925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1471142 c.1232G>T p.C411F Chr.17: 8110165-8110165 on Build GRCh38
1660502 c.1612A>C p.N538H Chr.17: 8110181-8110181 on Build GRCh38
6598722 c.1207C>G p.H403D Chr.17: 8110190-8110190 on Build GRCh38
4070860 c.1275C>A p.R425R Chr.17: 8110122-8110122 on Build GRCh38
4471307 c.1314C>T p.L438L Chr.17: 8109994-8109994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768625560 Chr.17: 8110140-8110140 on Build GRCh38
rs3027206 Chr.17: 8110156-8110156 on Build GRCh38
rs753269563 Chr.17: 8110068-8110068 on Build GRCh38
rs778696386 Chr.17: 8110091-8110091 on Build GRCh38
rs540629378 Chr.17: 8110177-8110177 on Build GRCh38
rs370970761 Chr.17: 8110037-8110037 on Build GRCh38
rs764522136 Chr.17: 8110077-8110077 on Build GRCh38
rs747998578 Chr.17: 8110176-8110176 on Build GRCh38
rs761072118 Chr.17: 8110075-8110075 on Build GRCh38
rs868428910 Chr.17: 8109994-8109994 on Build GRCh38
rs780048136 Chr.17: 8110009-8110009 on Build GRCh38
rs1017262034 Chr.17: 8109988-8109988 on Build GRCh38
rs746976183 Chr.17: 8110123-8110123 on Build GRCh38
rs746751980 Chr.17: 8110021-8110021 on Build GRCh38
rs761241768 Chr.17: 8110195-8110195 on Build GRCh38
rs900205585 Chr.17: 8109998-8109998 on Build GRCh38
rs763127321 Chr.17: 8110044-8110044 on Build GRCh38
rs776100693 Chr.17: 8110191-8110191 on Build GRCh38
rs994562882 Chr.17: 8110006-8110006 on Build GRCh38
rs58194728 Chr.17: 8110077-8110077 on Build GRCh38
rs374769325 Chr.17: 8110178-8110178 on Build GRCh38
rs767843896 Chr.17: 8110067-8110067 on Build GRCh38
rs754304050 Chr.17: 8110080-8110080 on Build GRCh38
rs960938579 Chr.17: 8110114-8110114 on Build GRCh38
rs866904860 Chr.17: 8110182-8110182 on Build GRCh38
rs373602905 Chr.17: 8110035-8110035 on Build GRCh38
rs759477802 Chr.17: 8110179-8110179 on Build GRCh38
rs768465921 Chr.17: 8110036-8110036 on Build GRCh38
rs139869133 Chr.17: 8110083-8110083 on Build GRCh38
rs753562107 Chr.17: 8109977-8109977 on Build GRCh38
rs906448267 Chr.17: 8110079-8110079 on Build GRCh38
rs991520978 Chr.17: 8110150-8110150 on Build GRCh38
rs771901360 Chr.17: 8109981-8109981 on Build GRCh38
rs779803421 Chr.17: 8110113-8110113 on Build GRCh38
rs771231164 Chr.17: 8110047-8110047 on Build GRCh38
rs774279430 Chr.17: 8110052-8110052 on Build GRCh38
rs769882705 Chr.17: 8110053-8110068 on Build GRCh38
rs757937060 Chr.17: 8110110-8110110 on Build GRCh38
rs772216649 Chr.17: 8110190-8110190 on Build GRCh38
rs765319587 Chr.17: 8110102-8110102 on Build GRCh38
rs749834593 Chr.17: 8110099-8110099 on Build GRCh38
rs759726176 Chr.17: 8110061-8110061 on Build GRCh38

View Full Product Details