Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99418927-99419186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH7
Entrez Gene ID
131
Gene Name
alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide
Gene Aliases
ADH4
UniGene
Hs.389
Gene Chromosome Location
Chr.4: 99412261-99435510 on Build GRCh38
Transcripts
NM_000673.4 NM_001166504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1485550 c.943C>T p.P315S Chr.4: 99419064-99419064 on Build GRCh38
1753441 c.877T>A p.S293T Chr.4: 99419106-99419106 on Build GRCh38
3598580 c.970G>A p.D324N Chr.4: 99419037-99419037 on Build GRCh38
4513661 c.946C>T p.P316S Chr.4: 99419061-99419061 on Build GRCh38
6512422 c.874G>T p.A292S Chr.4: 99419109-99419109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772199864 Chr.4: 99418955-99418955 on Build GRCh38
rs367907552 Chr.4: 99419013-99419013 on Build GRCh38
rs756177333 Chr.4: 99419145-99419145 on Build GRCh38
rs764589391 Chr.4: 99419123-99419123 on Build GRCh38
rs761334405 Chr.4: 99419016-99419016 on Build GRCh38
rs998511794 Chr.4: 99418951-99418951 on Build GRCh38
rs757607320 Chr.4: 99419135-99419135 on Build GRCh38
rs536459748 Chr.4: 99419073-99419073 on Build GRCh38
rs1035736686 Chr.4: 99418952-99418952 on Build GRCh38
rs984328720 Chr.4: 99419025-99419025 on Build GRCh38
rs777987364 Chr.4: 99418958-99418958 on Build GRCh38
rs758751969 Chr.4: 99419041-99419041 on Build GRCh38
rs922570035 Chr.4: 99419076-99419076 on Build GRCh38
rs568928644 Chr.4: 99419014-99419014 on Build GRCh38
rs999157109 Chr.4: 99419144-99419144 on Build GRCh38
rs780564087 Chr.4: 99419032-99419032 on Build GRCh38
rs762686196 Chr.4: 99419087-99419087 on Build GRCh38
rs758813649 Chr.4: 99419120-99419120 on Build GRCh38
rs751624401 Chr.4: 99419120-99419121 on Build GRCh38
rs1046200001 Chr.4: 99419102-99419102 on Build GRCh38
rs771242344 Chr.4: 99419067-99419067 on Build GRCh38
rs999607124 Chr.4: 99419117-99419117 on Build GRCh38
rs769763635 Chr.4: 99418997-99418997 on Build GRCh38
rs377606177 Chr.4: 99419147-99419147 on Build GRCh38
rs745937491 Chr.4: 99419139-99419139 on Build GRCh38
rs759583176 Chr.4: 99418993-99418993 on Build GRCh38
rs768247531 Chr.4: 99419158-99419158 on Build GRCh38
rs991610529 Chr.4: 99418964-99418964 on Build GRCh38
rs373645348 Chr.4: 99419099-99419099 on Build GRCh38
rs367823526 Chr.4: 99419105-99419105 on Build GRCh38
rs755023295 Chr.4: 99419135-99419137 on Build GRCh38
rs747120786 Chr.4: 99418959-99418959 on Build GRCh38
rs977044117 Chr.4: 99419055-99419055 on Build GRCh38
rs150551313 Chr.4: 99419115-99419115 on Build GRCh38
rs372287341 Chr.4: 99419134-99419134 on Build GRCh38
rs554686551 Chr.4: 99419119-99419119 on Build GRCh38
rs375718550 Chr.4: 99418967-99418967 on Build GRCh38
rs775429505 Chr.4: 99419084-99419084 on Build GRCh38
rs774102086 Chr.4: 99419091-99419091 on Build GRCh38
rs769630848 Chr.4: 99419080-99419080 on Build GRCh38
rs751965423 Chr.4: 99419128-99419128 on Build GRCh38
rs201018597 Chr.4: 99419004-99419004 on Build GRCh38
rs201128772 Chr.4: 99418960-99418960 on Build GRCh38
rs760167786 Chr.4: 99419114-99419114 on Build GRCh38
rs749296680 Chr.4: 99419154-99419154 on Build GRCh38
rs749932162 Chr.4: 99419020-99419020 on Build GRCh38
rs753010633 Chr.4: 99419035-99419035 on Build GRCh38
rs1049766738 Chr.4: 99419093-99419093 on Build GRCh38
rs777883734 Chr.4: 99419043-99419043 on Build GRCh38
rs767054835 Chr.4: 99419017-99419017 on Build GRCh38
rs141729668 Chr.4: 99419033-99419033 on Build GRCh38
rs761522191 Chr.4: 99419097-99419097 on Build GRCh38
rs148046430 Chr.4: 99419012-99419012 on Build GRCh38
rs915358164 Chr.4: 99419107-99419107 on Build GRCh38
rs747288350 Chr.4: 99419050-99419050 on Build GRCh38
rs370741519 Chr.4: 99419079-99419079 on Build GRCh38
rs376612553 Chr.4: 99419156-99419156 on Build GRCh38
rs768448563 Chr.4: 99419088-99419088 on Build GRCh38
rs775204347 Chr.4: 99419001-99419001 on Build GRCh38

View Full Product Details