Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72901151-72901376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A1
Entrez Gene ID
216
Gene Name
aldehyde dehydrogenase 1 family member A1
Gene Aliases
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1
UniGene
Hs.76392
Gene Chromosome Location
Chr.9: 72900662-72953317 on Build GRCh38
Transcripts
NM_000689.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
456057 c.1502C>T p.S501L Chr.9: 72901212-72901212 on Build GRCh38
5660690 c.1499A>T p.N500I Chr.9: 72901215-72901215 on Build GRCh38
7145307 c.1453G>A p.E485K Chr.9: 72901261-72901261 on Build GRCh38
4851698 c.1492C>G p.Q498E Chr.9: 72901222-72901222 on Build GRCh38
456058 c.1467C>G p.V489V Chr.9: 72901247-72901247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139965768 Chr.9: 72901250-72901250 on Build GRCh38
rs374263391 Chr.9: 72901309-72901309 on Build GRCh38
rs750848620 Chr.9: 72901308-72901308 on Build GRCh38
rs1006905867 Chr.9: 72901307-72901307 on Build GRCh38
rs965563862 Chr.9: 72901312-72901312 on Build GRCh38
rs764331830 Chr.9: 72901256-72901256 on Build GRCh38
rs907004264 Chr.9: 72901197-72901197 on Build GRCh38
rs897120108 Chr.9: 72901268-72901268 on Build GRCh38
rs772496943 Chr.9: 72901298-72901299 on Build GRCh38
rs767165050 Chr.9: 72901234-72901234 on Build GRCh38
rs8187997 Chr.9: 72901333-72901333 on Build GRCh38
rs758801059 Chr.9: 72901319-72901319 on Build GRCh38
rs1024605403 Chr.9: 72901332-72901332 on Build GRCh38
rs144085228 Chr.9: 72901209-72901209 on Build GRCh38
rs1044554187 Chr.9: 72901184-72901184 on Build GRCh38
rs780216064 Chr.9: 72901321-72901321 on Build GRCh38
rs757880822 Chr.9: 72901281-72901281 on Build GRCh38
rs368587531 Chr.9: 72901236-72901236 on Build GRCh38
rs995460983 Chr.9: 72901297-72901297 on Build GRCh38
rs747901481 Chr.9: 72901324-72901324 on Build GRCh38
rs765878116 Chr.9: 72901303-72901303 on Build GRCh38
rs371621448 Chr.9: 72901270-72901270 on Build GRCh38
rs1015191545 Chr.9: 72901242-72901242 on Build GRCh38

View Full Product Details