Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70702251-70702524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
-
UniGene
Hs.352156
Gene Chromosome Location
Chr.10: 70672518-70762441 on Build GRCh38
Transcripts
NM_080722.4 NM_139155.3 XM_011539300.2 XM_011539303.2 XM_011539305.2 XM_011539306.2 XM_011539307.2 XM_011539308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4144823 c.539C>A p.T180K Chr.10: 70702328-70702328 on Build GRCh38
4659474 c.661G>A p.G221R Chr.10: 70702450-70702450 on Build GRCh38
6537035 c.531C>G p.L177L Chr.10: 70702320-70702320 on Build GRCh38
5012170 c.543C>G p.D181E Chr.10: 70702332-70702332 on Build GRCh38
5806355 c.661G>T p.G221W Chr.10: 70702450-70702450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772708307 Chr.10: 70702401-70702401 on Build GRCh38
rs201260006 Chr.10: 70702383-70702383 on Build GRCh38
rs760334018 Chr.10: 70702316-70702316 on Build GRCh38
rs758366774 Chr.10: 70702362-70702362 on Build GRCh38
rs768168228 Chr.10: 70702496-70702496 on Build GRCh38
rs34022601 Chr.10: 70702324-70702324 on Build GRCh38
rs749658642 Chr.10: 70702290-70702290 on Build GRCh38
rs754041470 Chr.10: 70702449-70702449 on Build GRCh38
rs778561224 Chr.10: 70702391-70702393 on Build GRCh38
rs761019300 Chr.10: 70702429-70702429 on Build GRCh38
rs373981364 Chr.10: 70702393-70702393 on Build GRCh38
rs776039115 Chr.10: 70702497-70702497 on Build GRCh38
rs747772903 Chr.10: 70702500-70702500 on Build GRCh38
rs749512850 Chr.10: 70702374-70702374 on Build GRCh38
rs748317743 Chr.10: 70702285-70702285 on Build GRCh38
rs746245958 Chr.10: 70702457-70702457 on Build GRCh38
rs555523027 Chr.10: 70702436-70702436 on Build GRCh38
rs369388518 Chr.10: 70702338-70702338 on Build GRCh38
rs914521544 Chr.10: 70702442-70702442 on Build GRCh38
rs774899485 Chr.10: 70702297-70702297 on Build GRCh38
rs771257571 Chr.10: 70702291-70702291 on Build GRCh38
rs764128541 Chr.10: 70702447-70702447 on Build GRCh38
rs747506607 Chr.10: 70702485-70702485 on Build GRCh38
rs866420530 Chr.10: 70702330-70702330 on Build GRCh38
rs141259412 Chr.10: 70702415-70702415 on Build GRCh38
rs770233607 Chr.10: 70702288-70702289 on Build GRCh38
rs756187663 Chr.10: 70702437-70702437 on Build GRCh38
rs145852829 Chr.10: 70702328-70702328 on Build GRCh38
rs201809345 Chr.10: 70702325-70702325 on Build GRCh38
rs768113762 Chr.10: 70702406-70702406 on Build GRCh38
rs780698324 Chr.10: 70702478-70702478 on Build GRCh38
rs113554300 Chr.10: 70702364-70702364 on Build GRCh38
rs541878177 Chr.10: 70702476-70702476 on Build GRCh38
rs1023441102 Chr.10: 70702390-70702390 on Build GRCh38
rs368695287 Chr.10: 70702298-70702298 on Build GRCh38
rs376839956 Chr.10: 70702314-70702314 on Build GRCh38
rs775080060 Chr.10: 70702405-70702405 on Build GRCh38
rs141188417 Chr.10: 70702423-70702423 on Build GRCh38
rs375758998 Chr.10: 70702302-70702302 on Build GRCh38
rs373377233 Chr.10: 70702422-70702422 on Build GRCh38
rs750315083 Chr.10: 70702343-70702343 on Build GRCh38
rs111846456 Chr.10: 70702486-70702486 on Build GRCh38
rs898922441 Chr.10: 70702373-70702373 on Build GRCh38
rs775570196 Chr.10: 70702383-70702384 on Build GRCh38
rs761449372 Chr.10: 70702419-70702419 on Build GRCh38
rs199812543 Chr.10: 70702381-70702381 on Build GRCh38
rs778052718 Chr.10: 70702368-70702368 on Build GRCh38
rs746335847 Chr.10: 70702389-70702389 on Build GRCh38
rs371398989 Chr.10: 70702414-70702414 on Build GRCh38
rs376832712 Chr.10: 70702367-70702367 on Build GRCh38
rs372371243 Chr.10: 70702299-70702299 on Build GRCh38
rs549993695 Chr.10: 70702288-70702288 on Build GRCh38
rs35682622 Chr.10: 70702339-70702339 on Build GRCh38
rs376783762 Chr.10: 70702412-70702412 on Build GRCh38
rs879079326 Chr.10: 70702334-70702334 on Build GRCh38
rs776328715 Chr.10: 70702411-70702411 on Build GRCh38
rs1003049382 Chr.10: 70702340-70702340 on Build GRCh38
rs377541226 Chr.10: 70702450-70702450 on Build GRCh38
rs74797959 Chr.10: 70702363-70702363 on Build GRCh38
rs1053634748 Chr.10: 70702306-70702306 on Build GRCh38
rs201605109 Chr.10: 70702313-70702313 on Build GRCh38
rs777931308 Chr.10: 70702289-70702289 on Build GRCh38
rs779351289 Chr.10: 70702452-70702452 on Build GRCh38

View Full Product Details