Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149198150-149198393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM3
Entrez Gene ID
22885
Gene Name
actin binding LIM protein family member 3
Gene Aliases
HMFN1661
UniGene
Hs.49688
Gene Chromosome Location
Chr.5: 149141483-149260439 on Build GRCh38
Transcripts
NM_001301015.1 NM_001301018.1 NM_001301027.1 NM_001301028.1 NM_014945.3 XM_005268392.1 XM_005268393.1 XM_005268394.1 XM_005268395.1 XM_005268396.1 XM_011537609.2 XM_017009238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5060830 c.225G>T p.Q75H Chr.5: 149198292-149198292 on Build GRCh38
6591249 c.285G>A p.S95S Chr.5: 149198352-149198352 on Build GRCh38
736432 c.294C>A p.G98G Chr.5: 149198361-149198361 on Build GRCh38
1672028 c.221A>G p.Y74C Chr.5: 149198288-149198288 on Build GRCh38
3827364 c.305A>C p.H102P Chr.5: 149198372-149198372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199548691 Chr.5: 149198324-149198324 on Build GRCh38
rs769669191 Chr.5: 149198212-149198212 on Build GRCh38
rs768344927 Chr.5: 149198177-149198177 on Build GRCh38
rs373118040 Chr.5: 149198338-149198338 on Build GRCh38
rs943965898 Chr.5: 149198315-149198315 on Build GRCh38
rs748022848 Chr.5: 149198340-149198340 on Build GRCh38
rs749211850 Chr.5: 149198351-149198351 on Build GRCh38
rs769545470 Chr.5: 149198341-149198341 on Build GRCh38
rs977575822 Chr.5: 149198181-149198181 on Build GRCh38
rs770441625 Chr.5: 149198352-149198352 on Build GRCh38
rs762491848 Chr.5: 149198225-149198225 on Build GRCh38
rs757993871 Chr.5: 149198307-149198307 on Build GRCh38
rs377176079 Chr.5: 149198226-149198226 on Build GRCh38
rs780870461 Chr.5: 149198332-149198332 on Build GRCh38
rs754389834 Chr.5: 149198308-149198308 on Build GRCh38
rs530741758 Chr.5: 149198362-149198362 on Build GRCh38
rs765829047 Chr.5: 149198195-149198195 on Build GRCh38
rs780973202 Chr.5: 149198190-149198190 on Build GRCh38
rs759146275 Chr.5: 149198359-149198359 on Build GRCh38
rs753323555 Chr.5: 149198212-149198212 on Build GRCh38
rs750610592 Chr.5: 149198318-149198318 on Build GRCh38
rs953866469 Chr.5: 149198266-149198266 on Build GRCh38
rs868430620 Chr.5: 149198265-149198265 on Build GRCh38
rs778989066 Chr.5: 149198309-149198309 on Build GRCh38
rs746086610 Chr.5: 149198172-149198172 on Build GRCh38
rs375798677 Chr.5: 149198373-149198373 on Build GRCh38
rs761289040 Chr.5: 149198304-149198304 on Build GRCh38
rs752286038 Chr.5: 149198191-149198191 on Build GRCh38
rs773934073 Chr.5: 149198238-149198238 on Build GRCh38
rs764209198 Chr.5: 149198375-149198375 on Build GRCh38
rs773109048 Chr.5: 149198214-149198214 on Build GRCh38
rs145134862 Chr.5: 149198299-149198299 on Build GRCh38
rs548869504 Chr.5: 149198363-149198363 on Build GRCh38
rs767782062 Chr.5: 149198296-149198296 on Build GRCh38
rs974220651 Chr.5: 149198189-149198189 on Build GRCh38
rs759170287 Chr.5: 149198256-149198256 on Build GRCh38
rs375384469 Chr.5: 149198197-149198197 on Build GRCh38

View Full Product Details