Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38467293-38467542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP23
Entrez Gene ID
57636
Gene Name
Rho GTPase activating protein 23
Gene Aliases
-
UniGene
Hs.374446
Gene Chromosome Location
Chr.17: 38419286-38512392 on Build GRCh38
Transcripts
NM_001199417.1 XM_006721991.1 XM_006721992.3 XM_006721993.3 XM_006721994.3 XM_011525073.1 XM_011525074.2 XM_011525075.2 XM_011525076.2 XM_011525077.2 XM_017024902.1

SNP Detail

SNP ID Chromosome Location
rs945174130 Chr.17: 38467493-38467493 on Build GRCh38
rs116013904 Chr.17: 38467393-38467393 on Build GRCh38
rs192167502 Chr.17: 38467465-38467465 on Build GRCh38
rs138699241 Chr.17: 38467497-38467497 on Build GRCh38
rs913653351 Chr.17: 38467493-38467496 on Build GRCh38
rs776922140 Chr.17: 38467379-38467379 on Build GRCh38
rs1038881660 Chr.17: 38467327-38467327 on Build GRCh38
rs767724521 Chr.17: 38467382-38467383 on Build GRCh38
rs927977370 Chr.17: 38467369-38467369 on Build GRCh38
rs900802703 Chr.17: 38467479-38467479 on Build GRCh38
rs574499323 Chr.17: 38467428-38467428 on Build GRCh38
rs1014686394 Chr.17: 38467398-38467398 on Build GRCh38
rs994895856 Chr.17: 38467368-38467368 on Build GRCh38
rs145902466 Chr.17: 38467414-38467414 on Build GRCh38
rs980361100 Chr.17: 38467415-38467415 on Build GRCh38
rs745319805 Chr.17: 38467347-38467347 on Build GRCh38
rs902798278 Chr.17: 38467427-38467427 on Build GRCh38
rs8065549 Chr.17: 38467361-38467361 on Build GRCh38
rs187282060 Chr.17: 38467396-38467396 on Build GRCh38
rs938100301 Chr.17: 38467374-38467374 on Build GRCh38
rs911596944 Chr.17: 38467500-38467500 on Build GRCh38
rs1041144134 Chr.17: 38467461-38467461 on Build GRCh38
rs149190200 Chr.17: 38467399-38467399 on Build GRCh38
rs879257839 Chr.17: 38467356-38467356 on Build GRCh38
rs771432044 Chr.17: 38467371-38467371 on Build GRCh38
rs982558353 Chr.17: 38467363-38467363 on Build GRCh38

View Full Product Details